H2504
His-Leu
≥98% (TLC), suitable for ligand binding assays
Synonym(s):
histidylleucine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H20N4O3
CAS Number:
Molecular Weight:
268.31
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
His-Leu,
Assay
≥98% (TLC)
Quality Level
form
solid
technique(s)
ligand binding assay: suitable
color
white to off-white
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C(N)Cc1cnc[nH]1)C(O)=O
InChI
1S/C12H20N4O3/c1-7(2)3-10(12(18)19)16-11(17)9(13)4-8-5-14-6-15-8/h5-7,9-10H,3-4,13H2,1-2H3,(H,14,15)(H,16,17)(H,18,19)
InChI key
MMFKFJORZBJVNF-UHFFFAOYSA-N
Related Categories
Biochem/physiol Actions
Histidylleucine (His-Leu); histidylglycine (His-Gly) and histidylserine (His-Ser) are N-terminal imidazole containing dipeptides used to study binding of metals such as copper, nickel and zinc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Identification of enzymatically produced peptide fragments by liquid chromatography and mass spectrometry.
R Baranowski et al.
Analytical biochemistry, 121(1), 97-102 (1982-03-15)
S Ceccatelli et al.
Neuroendocrinology, 49(3), 309-323 (1989-03-01)
The distribution of several peptides, corticotropin-releasing factor (CRF), neurotensin (NT), enkephalin (ENK), vasoactive intestinal polypeptide (VIP), peptide histidine isoleucine (PHI), cholecystokinin (CCK), thyrotropin-releasing hormone (TRH) and galanin (GAL) was studied in detail with immunohistochemistry in the various subdivisions of the
K Iwatsuki et al.
Clinical and experimental pharmacology & physiology, 20(7-8), 501-507 (1993-07-01)
1. The effects of peptide histidine isoleucine (PHI) on pancreatic exocrine secretion were investigated in preparations of the isolated and blood-perfused dog pancreas as compared with those of vasoactive intestinal peptide (VIP), secretin and glucagon. 2. Each peptide tested was
R Reneland et al.
Scandinavian journal of clinical and laboratory investigation, 54(2), 105-111 (1994-04-01)
The importance of skeletal muscle for human haemodynamics and metabolism makes it a suitable sample tissue for the measurement of angiotensin converting enzyme (ACE; EC 3.4.15.1). A study of serum and muscle angiotensin-converting enzyme was performed in 50 essential-hypertensive subjects.
H Iwata et al.
Biotechnology progress, 7(5), 412-418 (1991-09-01)
An immobilized metal affinity (IMA) hollow-fiber membrane was prepared by radiation-induced graft polymerization of glycidyl methacrylate (GMA) onto a porous polyethylene hollow fiber, followed by chemical conversion of the produced epoxide group into an iminodiacetate (IDA) group and its chelation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service