Skip to Content
Merck
All Photos(6)

Key Documents

SAE0064

Sigma-Aldrich

Angiotensin Converting Enzyme-2, ACE2

Human recombinant, ≥1000000 U/mg, expressed in HEK 293 cells

Synonym(s):

ACE-related carboxypeptidase, ACE2, Angiotensin-converting enzyme homolog (ACEH), COVID-19 receptor, Coronavirus receptor, Metalloprotease MPROT15

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

human

Quality Level

100
200

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

specific activity

≥1000000 U/mg

mol wt

~85.9 kDa by SDS-PAGE

impurities

≤1 EU/μg protein Endotoxin

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... ACE2(59272)

General description

The gene for angiotensin converting enzyme-2 (ACE-2) is mapped to human chromosome Xp22.2. This protein is the first known human homologue of ACE. ACE2 has been identified from 5′ sequencing of a human heart failure ventricle cDNA library. It contains a signal peptide, one metalloprotease active site and a transmembrane domain. ACE-2 is a secreted and an integral membrane protein. It is expressed predominantly on the endothelium.

Application

Angiotensin Converting Enzyme-2 (ACE2) has been used in the ACE-2 inhibition assays for inhibitor selectivity studies. It has also been used for identifying potent ACE2-blocking monoclonal antibodies in an on-chip assay and in human-ACE2-blocking assay using a biolayer interferometry biosensor. This approach serves for rational vaccine designing and for the selection of robust immunotherapeutic agents against the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection.

Biochem/physiol Actions

Recombinant human Angiotensin Converting Enzyme-2 (ACE-2) is expressed in human HEK 293 cells as a C-terminally flag and histidine-tagged glycoprotein with a calculated molecular mass of 85.9 kDa (amino acids Gln18-Ser740, with a C-terminal 10-His tag). The DTT-reduced protein migrates as a 90-120 kDa polypeptide on SDS-PAGE due to glycosylation. This protein is manufactured in human cells, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports higher specific activity of the protein.
The angiotensin converting enzyme-2 (ACE-2) functions as carboxydipeptidase. The central role of ACE-2, is to counter ACE activity by reducing the bioavailability of angiotensin (Ang)-II and increasing the Ang(1-7) formation. ACE-2 is a part of the renin-angiotensin system (RAS). Many researches show that Ace-2 ensures the protection of peripheral tissues and might be efficient to treat RAS-related diseases. Also, an imbalance in Ace-2/Ang-(1-7) and ACE/Ang-II axes is important for the onset of cardiovascular diseases. Severe acute respiratory syndrome (SARS) and human coronavirus (HCoV)-NL63 infection viruses has been shown to use its surface protein spike (glycoprotein) to bind to human ACE-2 receptor. The S protein is cleaved into subunits, S1 and S2 during the COVID-19 infection. S1 contains the receptor binding domain (RBD) which enables coronaviruses to bind to the peptidase domain (PD) of ACE2. Thus, ACE2 has become a high focus research target for COVID-19 infection.

Unit Definition

One unit is defined as the amount of enzyme required to cleave 1 picomole of the fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH in one minute, in 37 °C, pH 7.5.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potently neutralizing and protective human antibodies against SARS-CoV-2
Zost SJ, et al.
Nature, 1-7 (2020)
Huijing Xia et al.
Current hypertension reports, 12(3), 170-175 (2010-04-29)
Angiotensin-converting enzyme 2 (ACE2) is a new component of the renin-angiotensin system (RAS). Accumulating evidence shows that ACE2 provides protective effects in peripheral tissues and has great potential for the treatment of RAS-related diseases. The role of ACE2 in the
Rapid isolation and profiling of a diverse panel of human monoclonal antibodies targeting the SARS-CoV-2 spike protein
<BIG>Zost SJ, et al. </BIG>
Nature Medicine (2020)
Yao-Ling Wang et al.
Journal of medicinal chemistry, 62(15), 7160-7184 (2019-07-04)
The emergence and spread of bacterial pathogens acquired metallo-β-lactamase (MBL) and serine-β-lactamase (SBL) medicated β-lactam resistance gives rise to an urgent need for the development of new dual-action MBL/SBL inhibitors. Application of a pharmacophore fusion strategy led to the identification
Fang Li
Journal of virology, 89(4), 1954-1964 (2014-11-28)
Receptor recognition by viruses is the first and essential step of viral infections of host cells. It is an important determinant of viral host range and cross-species infection and a primary target for antiviral intervention. Coronaviruses recognize a variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service