Skip to Content
Merck
All Photos(1)

Key Documents

94403

Sigma-Aldrich

Hoechst 33258 solution

1 mg/mL in H2O, ≥98.0% (HPLC)

Synonym(s):

bisBenzimide H 33258 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N6O · 3HCl
CAS Number:
Molecular Weight:
533.88
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

form

liquid

concentration

1 mg/mL in H2O

solubility

DMF: soluble
H2O: soluble

fluorescence

λex 355 nm; λem 465 nm in TE buffer; DNA

storage temp.

2-8°C

SMILES string

Cl.Cl.Cl.CN1CCN(CC1)c2ccc3[nH]c(nc3c2)-c4ccc5nc([nH]c5c4)-c6ccc(O)cc6

InChI

1S/C25H24N6O.3ClH/c1-30-10-12-31(13-11-30)18-5-9-21-23(15-18)29-25(27-21)17-4-8-20-22(14-17)28-24(26-20)16-2-6-19(32)7-3-16;;;/h2-9,14-15,32H,10-13H2,1H3,(H,26,28)(H,27,29);3*1H

InChI key

SMNPLAKEGAEPJD-UHFFFAOYSA-N

General description

Hoechst 33258, also known as bisbenzimide, is a fluorescent blue dye. [1]It is categorized as a chromosomal stain that binds to the minor groove of the DNA double helix recognizing a run of four A-T base pairs.[2]

Application

Hoechst 33258 is used to estimate DNA concentration in samples by fluorometry which is better than spectrophotometry, allowing the determination of nanogram quantities of DNA.[3] Also, Hoechst 33258 is suitable to detect mycoplasma contamination in cell cultures due to its high fluorescence ability.[4]

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael R Green et al.
Cold Spring Harbor protocols, 2017(5) (2017-05-04)
Measuring the concentration of DNA using fluorometry is more sensitive than spectrophotometry, allowing the detection of nanogram quantities of DNA. In this assay, DNA preparations of known and unknown concentrations are incubated with the fluorochrome Hoechst 33258. Absorption values for
Natalia Busto et al.
The journal of physical chemistry. B, 119(13), 4575-4581 (2015-03-12)
The functionality of the bisbenzimide Hoechst 33258 in solution has been largely exploited in the quantification of DNA. Understanding of its behavior is essential to promote its widespread application and learning of biological processes. A detailed study of the dimerization
In situ detection of mycoplasma contamination in cell cultures by fluorescent Hoechst 33258 stain.
T R Chen
Experimental cell research, 104(2), 255-262 (1977-02-01)
Cornelia Fritsch et al.
PLoS genetics, 15(7), e1008269-e1008269 (2019-07-13)
Development of eye tissue is initiated by a conserved set of transcription factors termed retinal determination network (RDN). In the fruit fly Drosophila melanogaster, the zinc-finger transcription factor Glass acts directly downstream of the RDN to control identity of photoreceptor
A Fede et al.
Structure (London, England : 1993), 1(3), 177-186 (1993-11-15)
The chromosomal stain, Hoechst 33258, binds to the minor groove of the DNA double helix and specifically recognizes a run of four A-T base pairs. Extensive biochemical and biophysical studies have been aimed at understanding the binding of the dye

Protocols

Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.

Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.

Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.

Cultivate ReNcell® human neural stem cells in 3D hydrogels for high-throughput screening using the TrueGel3D® HTS Hydrogel Plate with this protocol.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service