Skip to Content
Merck
All Photos(1)

Key Documents

63493

Sigma-Aldrich

Hoechst 34580

Trihydrochloride salt, ≥98.0% (HPLC)

Synonym(s):

N,N-Dimethyl-4-[5-(4-methyl-1-piperazinyl)[2,5′-bi-1H-benzimidazol]-2′-yl]benzenamine trihydrochloride, HOE 34580

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29N7 · 3HCl
CAS Number:
Molecular Weight:
560.95
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98.0% (HPLC)

quality

Trihydrochloride salt

solubility

H2O: 1 mg/mL, clear, slightly yellow to deep greenish-yellow

fluorescence

λex 357 nm; λem 490 nm±10 nm in H2O (free dye)

storage temp.

−20°C

SMILES string

Cl.Cl.Cl.CN1CCN(CC1)c2ccc3nc([nH]c3c2)-c4ccc5nc([nH]c5c4)-c6ccc(cc6)N(C)C

InChI

1S/C27H29N7.3ClH/c1-32(2)20-7-4-18(5-8-20)26-28-22-10-6-19(16-24(22)30-26)27-29-23-11-9-21(17-25(23)31-27)34-14-12-33(3)13-15-34;;;/h4-11,16-17H,12-15H2,1-3H3,(H,28,30)(H,29,31);3*1H

InChI key

IPMZLKUUQSFUTA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alwin de Jong et al.
Frontiers in cardiovascular medicine, 7, 604821-604821 (2021-02-16)
Background: Inflammatory stimuli induced by NF-kB drive atherosclerotic lesion formation. The epigenetic P300/CBP associated factor (PCAF) post-transcriptionally acetylates FoxP3, which is required for regulatory T-cell (Treg) differentiation and immune modulation. We hypothesize that PCAF deficiency affects atherosclerosis via regulation of
Irena Roci et al.
Cell cycle (Georgetown, Tex.), 19(20), 2676-2684 (2020-10-06)
Proliferating cells must synthesize a wide variety of macromolecules while progressing through the cell cycle, but the coordination between cell cycle progression and cellular metabolism is still poorly understood. To identify metabolic processes that oscillate over the cell cycle, we
Leonie Rosenberger et al.
Scientific reports, 9(1), 663-663 (2019-01-27)
Recently, exosomes secreted by menstrual mesenchymal stem cells have been identified as inhibitory agents of tumor angiogenesis and modulators of the tumor cell secretome in prostate and breast cancer. However, their direct effect on endothelial cells and paracrine mediators have
Koos Rooijers et al.
Nature biotechnology, 37(7), 766-772 (2019-06-19)
Protein-DNA interactions are critical to the regulation of gene expression, but it remains challenging to define how cell-to-cell heterogeneity in protein-DNA binding influences gene expression variability. Here we report a method for the simultaneous quantification of protein-DNA contacts by combining
Eveline A C Goossens et al.
International journal of molecular sciences, 21(10) (2020-05-21)
Myostatin is a negative regulator of muscle cell growth and proliferation. Furthermore, myostatin directly affects the expression of 14q32 microRNAs by binding the 14q32 locus. Direct inhibition of 14q32 microRNA miR-495-3p decreased postinterventional restenosis via inhibition of both vascular smooth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service