Skip to Content
Merck
All Photos(1)

Key Documents

43815

Sigma-Aldrich

DL-Dithiothreitol

BioUltra, for molecular biology, ≥99.5% (RT)

Synonym(s):

(±)-Dithiothreitol, rac-Dithiothreitol, Dithiothreitol, threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719757
EC Number:
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.5% (RT)

form

powder

reaction suitability

reagent type: reductant

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.3% 4,5-dihydroxy-1,2-dithiane

pH

4.0-6.5 (25 °C, 0.1 M in H2O)

mp

41-44 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.400
λ: 280 nm Amax: 0.100

SMILES string

O[C@H](CS)[C@H](O)CS

storage temp.

2-8°C

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dithiothreitol (DTT) is a thiol-reducing reagent that quantitatively reduces disulfide (SH) groups in biological molecules. It specifically targets the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and less toxic than 2-mercaptoethanol. Typically, a seven-fold lower concentration of DTT (100 mM) is more efficient than that of 2-mercaptoethanol (5% v/v, 700 mM).

Application

DTT has been used:
  • as an enzyme stabilizer to maintain exebacase stability
  • for peptide extraction and protein denaturation
  • in sample preparation and reversal of formaldehyde crosslinks before mass spectrometry
  • as a buffer component to extract synaptogliosome
An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

Biochem/physiol Actions

Dithiothreitol (DTT) is broadly used in chemical peptide synthesis and biochemical preparations of thiol proteins. Additionally, studies on protein folding and enzyme activity in protein chemistry make use of DTT. This reagent explicitly mediates the thiol-disulfide exchange reaction to completely reduce the intra-or between sub-atomic disulfide bonds in biomolecules. This reaction brings about the arrangement of thiols and the cyclic disulfide of DTT.

Features and Benefits

  • BioUltra grade powder suitable for molecular biology
  • DNase, RNase, phosphatase, and protease-free insoluble matter, passes the filter test
  • Stringently tested and free from trace metals

Other Notes

For additional information on our range of Biochemicals, please complete this form.
Isolation of RNA using guanidinium salts. Inclusion of a reductant enhances denaturation by breaking intramolecular protein disulfide bonds.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunoprecipitation of Ribosome-Bound mRNAs from Astrocytic Perisynaptic Processes of the Mouse Hippocampus
Mazare N, et al.
STAR protocols (2022)
Generation of cell-type-specific proteomes of neurodevelopment from human cerebral organoids
Melliou S and Diamandis P
STAR protocols (2022)
Jun T Oh et al.
Antimicrobial agents and chemotherapy, 65(7), e0258720-e0258720 (2021-04-28)
Exebacase (CF-301) belongs to a new class of protein-based antibacterial agents, known as lysins (peptidoglycan hydrolases). Exebacase, a novel lysin with antistaphylococcal activity, is in phase 3 of clinical development. To advance into the clinic, it was necessary to develop
John C Lukesh et al.
Journal of the American Chemical Society, 134(9), 4057-4059 (2012-02-23)
Dithiothreitol (DTT) is the standard reagent for reducing disulfide bonds between and within biological molecules. At neutral pH, however, >99% of DTT thiol groups are protonated and thus unreactive. Herein, we report on (2S)-2-amino-1,4-dimercaptobutane (dithiobutylamine or DTBA), a dithiol that
Isolation of RNA using guanidinium salts.
R J MacDonald et al.
Methods in enzymology, 152, 219-227 (1987-01-01)

Articles

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Understand how mRNA vaccines induce immunity. and read how synthetic mRNA is prepared for vaccine immunogens and other biopharmaceuticals. Find reagents for synthesis of mRNA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service