Skip to Content
Merck
All Photos(4)

Key Documents

D9163

Sigma-Aldrich

DL-Dithiothreitol

suitable for electrophoresis, ≥99%

Synonym(s):

threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719757
EC Number:
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NB.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99%

form

powder

reaction suitability

reagent type: reductant

technique(s)

electrophoresis: suitable

mp

40-44 °C
41-44 °C (lit.)

solubility

H2O: 50 mg/mL, clear, colorless to very faintly yellow

cation traces

heavy metals (as Pb): ≤10 ppm

absorption

≤0.04 at 280 at 0.02 M

storage temp.

2-8°C

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

Analysis Note

Tested for use in reduced SDS-PAGE

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liisa Arike et al.
Cell reports, 30(4), 1077-1087 (2020-01-30)
The gastrointestinal tract is covered by a single layer of epithelial cells that, together with the mucus layers, protect the underlying tissue from microbial invasion. The epithelium has one of the highest turnover rates in the body. Using stable isotope
Jon Landa et al.
Neurology(R) neuroimmunology & neuroinflammation, 8(1) (2020-11-05)
To describe the clinical syndrome of 4 new patients with seizure-related 6 homolog like 2 antibodies (SEZ6L2-abs), study the antibody characteristics, and evaluate their effects on neuronal cultures. SEZ6L2-abs were initially identified in serum and CSF of a patient with
Baoyi Zhu et al.
Journal of cellular physiology, 235(10), 7370-7382 (2020-02-11)
Smooth muscle cells (SMCs) are characterized by a high degree of phenotypic plasticity. Contractile differentiation is governed by myocardin-related transcription factors (MRTFs), in particular myocardin (MYOCD), and when their drive is lost, the cells become proliferative and synthetic with an
Florentina Negoita et al.
Biochemical and biophysical research communications, 508(3), 775-779 (2018-12-12)
Dysregulation of autophagy has been observed in obesity and type 2 diabetes. Salt-inducible kinase 2 (SIK2), a member of the AMPK-related kinase family, is downregulated in adipocytes from obese or insulin resistant individuals and was previously demonstrated to regulate autophagy
Xueqian Liu et al.
The Journal of biological chemistry, 288(10), 7230-7240 (2013-01-25)
In Pichia pastoris, the peroxisomal targeting signal 2 (PTS2)-dependent peroxisomal matrix protein import pathway requires the receptor, Pex7, and its co-receptor Pex20. A conserved lysine (Lys(19)) near the N terminus of Pex20 is required for its polyubiquitination and proteasomal degradation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service