Skip to Content
Merck
All Photos(1)

Key Documents

05129

Sigma-Aldrich

L-Alanine

BioUltra, ≥99.5% (NT)

Synonym(s):

(S)-2-Aminopropionic acid, L-α-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
Beilstein:
1720248
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

Quality Level

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D +14.0±1.0°, c = 5% in 5 M HCl

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.05%

loss

≤0.05% loss on drying, 20 °C (HV)

color

colorless to white

pH

5.5-7.0 (25 °C, 1 M in H2O)

mp

314.5 °C

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤100 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.03
λ: 280 nm Amax: ≤0.02

application(s)

detection

SMILES string

C[C@H](N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1

InChI key

QNAYBMKLOCPYGJ-REOHCLBHSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

Application


  • A Color Indicator Based on 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium Bromide (MTT) and a Biodegradable Poly(ester amide) for Detecting Bacterial Contamination.: This study developed a biodegradable poly(ester amide) containing L-Alanine used with MTT for the detection of bacterial contamination, highlighting its potential in health-related applications (Franco et al., 2024).

  • Randomized Trial to Assess the Safety and Tolerability of Daily Intake of an Allulose Amino Acid-Based Hydration Beverage in Men and Women.: Investigates the impact of a daily hydration beverage supplemented with L-Alanine among other amino acids on hydration and nutrient uptake in humans, emphasizing its safety and efficacy (Bloomer et al., 2024).

  • Comparison of methods for the NMR measurement of motionally averaged dipolar couplings.: This study utilizes L-Alanine samples to compare NMR methods for measuring dipolar couplings, contributing to the improvement of NMR spectroscopy techniques (Southern et al., 2024).

Biochem/physiol Actions

Alanine is a nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions. The amino group of glutamate is transferred to pyruvate via alanine aminotransferase, forming alanine and α-ketoglutarate. The alanine is passed into the blood and transported to the liver. This reaction is reversed in the liver where pyruvate can be used in gluconeogenesis to form glucose, which may return to other tissues through the circulatory system. Increased alanine levels correlate with higher blood pressure, energy intake, cholesterol levels, and body mass index.

Other Notes

Postulated salting-in effect on proteins; Growth requirement of a variety of microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

CRC Handbook of Microbiology, 4, 1-1 (1974)
Theory of protein solubility.
T Arakawa et al.
Methods in enzymology, 114, 49-77 (1985-01-01)
J M Rhoads et al.
Journal of pediatric gastroenterology and nutrition, 9(2), 225-231 (1989-08-01)
We studied sodium-dependent uptake of L-alanine into small intestinal brush border membrane vesicles (BBMV) isolated from piglets 40 h after infection with transmissible gastroenteritis (TGE) virus. Vesicles from TGE-infected pigs and uninfected litter-mate controls showed comparable degrees of enrichment and
Lillian L Siu et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(19), 2477-2484 (2013-05-22)
The antiepidermal growth factor receptor monoclonal antibody cetuximab has improved survival in patients with metastatic, chemotherapy-refractory, wild-type K-RAS colorectal cancer. The addition of brivanib, a tyrosine kinase inhibitor targeting vascular endothelial growth factor receptor and fibroblast growth factor receptor, to
Rachelle S Doody et al.
The New England journal of medicine, 369(4), 341-350 (2013-07-26)
Alzheimer's disease is characterized by the presence of cortical amyloid-beta (Aβ) protein plaques, which result from the sequential action of β-secretase and γ-secretase on amyloid precursor protein. Semagacestat is a small-molecule γ-secretase inhibitor that was developed as a potential treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service