Merck
  • Phase III randomized, placebo-controlled study of cetuximab plus brivanib alaninate versus cetuximab plus placebo in patients with metastatic, chemotherapy-refractory, wild-type K-RAS colorectal carcinoma: the NCIC Clinical Trials Group and AGITG CO.20 Trial.
Phase III randomized, placebo-controlled study of cetuximab plus brivanib alaninate versus cetuximab plus placebo in patients with metastatic, chemotherapy-refractory, wild-type K-RAS colorectal carcinoma: the NCIC Clinical Trials Group and AGITG CO.20 Trial.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-05-22)
Lillian L Siu, Jeremy D Shapiro, Derek J Jonker, Chris S Karapetis, John R Zalcberg, John Simes, Felix Couture, Malcolm J Moore, Timothy J Price, Jehan Siddiqui, Louise M Nott, Danielle Charpentier, Winston Liauw, Michael B Sawyer, Michael Jefford, Nadine M Magoski, Andrew Haydon, Ian Walters, Jolie Ringash, Dongsheng Tu, Chris J O'Callaghan
ABSTRACT
The antiepidermal growth factor receptor monoclonal antibody cetuximab has improved survival in patients with metastatic, chemotherapy-refractory, wild-type K-RAS colorectal cancer. The addition of brivanib, a tyrosine kinase inhibitor targeting vascular endothelial growth factor receptor and fibroblast growth factor receptor, to cetuximab has shown encouraging early clinical activity. Patients with metastatic colorectal cancer previously treated with combination chemotherapy were randomly assigned 1:1 to receive cetuximab 400 mg/m(2) intravenous loading dose followed by weekly maintenance of 250 mg/m(2) plus either brivanib 800 mg orally daily (arm A) or placebo (arm B). The primary end point was overall survival (OS). A total of 750 patients were randomly assigned (376 in arm A and 374 in arm B). Median OS in the intent-to-treat population was 8.8 months in arm A and 8.1 months in arm B (hazard ratio [HR], 0.88; 95% CI, 0.74 to 1.03; P = .12). Median progression-free survival (PFS) was 5.0 months in arm A and 3.4 months in arm B (HR, 0.72; 95% CI, 0.62 to 0.84; P < .001). Partial responses observed (13.6% v 7.2%; P = .004) were higher in arm A. Incidence of any grade ≥ 3 adverse events was 78% in arm A and 53% in arm B. Fewer patients received ≥ 90% dose-intensity of both cetuximab (57% v 83%) and brivanib/placebo (48% v 87%) in arm A versus arm B, respectively. Despite positive effects on PFS and objective response, cetuximab plus brivanib increased toxicity and did not significantly improve OS in patients with metastatic, chemotherapy-refractory, wild-type K-RAS colorectal cancer.
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, ≥99%
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C