1250
L-Alanine
≥98% dry basis (titration)
Synonym(s):
L-Alanine, Ala
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
L-Alanine, A non-essential amino acid.
Quality Level
Assay
≥98% dry basis (titration)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
water: 5 mg/mL
cation traces
heavy metals: ≤10 ppm
shipped in
ambient
storage temp.
15-25°C
InChI
1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)
InChI key
QNAYBMKLOCPYGJ-UHFFFAOYSA-N
General description
A non-essential amino acid.
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstition, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of cell science, 127(Pt 11), 2554-2564 (2014-04-04)
Bromodomain-containing protein 2 (Brd2) is a BET family chromatin adaptor required for expression of cell-cycle-associated genes and therefore involved in cell cycle progression. Brd2 is expressed in proliferating neuronal progenitors, displays cell-cycle-stimulating activity and, when overexpressed, impairs neuronal differentiation. Paradoxically
PloS one, 5(6), e11352-e11352 (2010-07-03)
TIA-1-related (TIAR) protein is a shuttling RNA-binding protein involved in several steps of RNA metabolism. While in the nucleus TIAR participates to alternative splicing events, in the cytoplasm TIAR acts as a translational repressor on specific transcripts such as those
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 37(4), 515-528 (1989-04-01)
We studied the subcellular localization of two major secretory products of adult rat submandibular gland (RSMG), blood group A-reactive mucin glycoprotein and glutamine/glutamic acid-rich protein (GRP), by light and electron microscopic immunocytochemistry. The structure of the major neutral oligosaccharide of
Nematocyst types and venom effects of Aurelia aurita and Velella velella from the Mediterranean Sea.
Toxicon : official journal of the International Society on Toxinology, 175, 57-63 (2020-02-15)
Natural substances produced by venomous marine organisms are thought to be possible sources of useful compounds and new drugs having the potential to open new ways for pharmacology, nutrition and environmental applications. In this framework, cnidarians are very interesting being
Biomedical optics express, 8(2), 1250-1256 (2017-03-09)
This study presents the screening of dengue virus (DENV) infection in human blood sera based on lactate concentration using Raman spectroscopy. A total of 70 samples, 50 from confirmed DENV infected patients and 20 from healthy volunteers have been used
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service