A1522
Aluminum oxide
Type WN-6, Neutral, Activity Grade Super I
Synonym(s):
Alumina
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
SB.52
Recommended Products
Quality Level
type
Type WN-6, Neutral
form
powder
mp
2040 °C (lit.)
SMILES string
O=[Al]O[Al]=O
InChI
1S/2Al.3O
InChI key
TWNQGVIAIRXVLR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Aluminum oxide (alumina, Al2O3), a metal oxide, is a common raw material for manufacturing ceramics. It is a good electrical insulator that shows stability at high temperature and resistance to chemicals. Alumina is an important source for aluminum and can be prepared from high-alumina ladle furnace refining slag. It can enhance the reaction rate of aldose to ketose conversion by Lobry de Bruyn–Alberda van Ekenstein transformation. It is a key constituent of NOx storage catalysts (NSC), which are employed to get rid of nitrogen oxides from the exhaust gas of lean burn engines. Its surface passivation characteristics on crystalline silicon makes it a vaiable candidate in silicon solar cell production.
Application
Aluminum oxide was used in the disposable alumina columns, to determine the adenylyl cyclase (AC) activity during the interaction of 2,3-O-(2,4,6-trinitrophenyl)-substituted nucleoside 5-triphosphates (TNP-NTPs)with VC1:IIC2 (purified catalytic subunits of membranous AC). It may be used as a dopant to produce optical fiber amplifiers and high-power fiber lasers.
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Srividya Suryanarayana et al.
The Journal of pharmacology and experimental therapeutics, 330(3), 687-695 (2009-06-06)
Adenylyl cyclases (ACs) catalyze the conversion of ATP into the second messenger cAMP and play a key role in signal transduction. In a recent study (Mol Pharmacol 70:878-886, 2006), we reported that 2',3'-O-(2,4,6-trinitrophenyl)-substituted nucleoside 5'-triphosphates (TNP-NTPs) are potent inhibitors (K(i)
High contrast laser marking of alumina.
Penide J, et al.
Applied Surface Science, 336, 118-128 (2015)
Aldose-ketose interconversion in pyridine in the presence of aluminium oxide.
Ekeberg D, et al.
Carbohydrate Research, 342(14), 1992-1997 (2007)
Nonlinear relaxational polarization in aluminum oxide.
Kankate L, et al.
IEEE Transactions on Dielectrics and Electrical Insulation, 22(2), 1220-1231 (2015)
Alumina extraction from high-alumina ladle furnace refining slag.
Zhou Y, et al.
Hydrometallurgy, 140, 14-19 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service