Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum oxide
Synonym(s):
Alumina
Linear Formula:
Al
2
O
3
CAS Number:
1344-28-1
Molecular Weight:
101.96
EC Number:
215-691-6
Products
Brand
Sigma-Aldrich (24)
Supelco (1)
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Application
Pore Size (Å)
Technique
Filter Products
Showing 1-20 of 25
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
199443
Aluminum oxide,
activated, basic, Brockmann I
Expand
View Pricing
199974
Aluminum oxide,
activated, neutral, Brockmann I
Expand
View Pricing
229423
Aluminum oxide,
powder, 99.99% trace metals basis
Expand
View Pricing
199966
Aluminum oxide,
activated, acidic, Brockmann I
Expand
View Pricing
265497
Aluminum oxide,
powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Expand
View Pricing
544833
Aluminum oxide,
nanopowder, <50 nm particle size (TEM)
Expand
View Pricing
702129
Aluminum oxide,
nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Expand
View Pricing
414069
Aluminum oxide,
pellets, 3 mm
Expand
View Pricing
769290
Aluminum oxide,
powder, ultra dry
Expand
View Pricing
234745
Aluminum oxide,
Corundum, α-phase, -100 mesh
Expand
View Pricing
718475
Aluminum oxide,
nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Expand
View Pricing
342688
Aluminum oxide,
fused, powder, primarily
α
-phase, -325 mesh
Expand
View Pricing
934054
Aluminum oxide,
gamma phase, nanopowder, 5 nm avg. part. size, 99.95% trace metals basis
Expand
View Pricing
202606
Aluminum oxide,
99.997% trace metals basis
Expand
View Pricing
267740
Aluminum oxide,
pore size 58 Å, ~150 mesh
Expand
View Pricing
642991
Aluminum oxide,
nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H
2
O
Expand
View Pricing
A1522
Aluminum oxide,
Type WN-6, Neutral, Activity Grade Super I
Expand
View Pricing
1.01095
Aluminum oxide,
(anhydrous (γ-aluminia)), Technipur
®
Expand
View Pricing
342653
Aluminum oxide,
fused, powder, primarily
α
-phase, 100-200 mesh
Expand
View Pricing
934062
Aluminum oxide,
alpha phase, nanopowder, 135 nm avg. part. size, 99.9% trace metals basis
Expand
View Pricing
Page 1 of 2
Page 1 of 2