Skip to Content
Merck
All Photos(2)

Key Documents

544833

Sigma-Aldrich

Aluminum oxide

nanopowder, <50 nm particle size (TEM)

Synonym(s):

Gamma Al2O3, Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
NA.23

description

gamma phase

Quality Level

form

nanopowder

surface area

>40 m2/g , BET

particle size

<50 nm (TEM)

mp

2040 °C (lit.)

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum oxide (Al2O3) based nanopowder is a metal oxide based nanosized material that can be used in a variety of industrial applications. It can be prepared by the hydrolysis of the alkoxide of Al2O3 and calcination of particles in the presence of stabilizing agents.

Application

Al2O3 nanopowder can be used for a wide range of applications, such as ceramics, aerospace, wastewater treatment, abrasives, paints, catalysts, and flooring materials.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nanotechnology in high performance paint coatings
Khanna AS
Asian Journal of Experimental Sciences, 21(2), 25-32 (2008)
Burning of nano-aluminized composite rocket propellants
De Luca LT, et al.
Combustion, Explosion and Shock Waves, 41(6), 680-692 (2005)
A study on the friction and wear behavior of PTFE filled with alumina nanoparticles
Sawyer WG, et al.
Wear, 254(5-6), 573-580 (2003)
Structure and properties of Zn-Al-Cu alloy reinforced with alumina particles
Martinez-Flores E, et al.
Materials & Design, 24(4), 281-286 (2003)
Structural transformations of alumina by high energy ball milling
ZielinAski PA
Journal of Materials Research, 8(11), 2985-2992 (1993)

Articles

Composite materials with micron scale reinforcements offer tailored properties for various applications.

Composite materials with micron scale reinforcements offer tailored properties for various applications.

Composite materials with micron scale reinforcements offer tailored properties for various applications.

Composite materials with micron scale reinforcements offer tailored properties for various applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service