Skip to Content
Merck
All Photos(3)

Key Documents

799351

Sigma-Aldrich

Sodium nitrate

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

pH

5.5-8 (20 °C, 50 g/L)

mp

306 °C (dec.) (lit.)

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chengjin Wang et al.
Environmental science & technology, 54(3), 1929-1937 (2019-12-28)
The hydroxyl radical (•OH) scavenging capacity is a useful parameter for the design and operation of an advanced oxidation process (AOP) in water treatment. The scavenging capacity may change with time, and it would be useful to continuously measure this
Adriana Ibarra-Hernández et al.
Nanomaterials (Basel, Switzerland), 8(2) (2018-02-14)
Graphite oxide is synthesized via oxidation reaction using oxidant compounds that have lattice defects by the incorporation of unlike functional groups. Herein, we report the synthesis of the graphite oxide with diverse surface oxygen content through three (B, C, D)
Yasser Vasseghian et al.
Food research international (Ottawa, Ont.), 137, 109557-109557 (2020-11-26)
In the current study, the Malathion decontamination by the aid of the UV/ferrous-activated persulfate (PS) was investigated and the effects of pH, persulfate (PS) concentration, ferrous concentration, Malathion concentration, and different inorganic ions were evaluated. Also, the Adaptive Neuro-Fuzzy Inference
Nan Wang et al.
Small (Weinheim an der Bergstrasse, Germany), e1801346-e1801346 (2018-06-22)
Due to substantial phonon scattering induced by various structural defects, the in-plane thermal conductivity (K) of graphene films (GFs) is still inferior to the commercial pyrolytic graphite sheet (PGS). Here, the problem is solved by engineering the structures of GFs
Xiangyu Zhu et al.
Nanoscale, 12(8), 4882-4894 (2020-01-10)
A deep understanding of the shaping technique is urgently required to precisely tailor the pore structure of a graphene aerogel (GA) in order to fit versatile application backgrounds. In the present study, the microstructure and properties of GA were regulated

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service