Skip to Content
Merck
All Photos(1)

Key Documents

44939

Sigma-Aldrich

Iron(II) chloride tetrahydrate

puriss. p.a., ≥99.0% (RT)

Synonym(s):

Ferrous chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2 · 4H2O
CAS Number:
Molecular Weight:
198.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (RT)
form:
powder or crystals

vapor pressure

10 mmHg ( 693 °C)

Quality Level

grade

puriss. p.a.

Assay

≥99.0% (RT)

form

powder or crystals

impurities

≤0.001% total nitrogen (N)

pH

2.5 (20 °C, 100 g/L)

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cr: ≤10 mg/kg
Cu: ≤5 mg/kg
Fe3+: ≤2000 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤10 mg/kg
Zn: ≤20 mg/kg

SMILES string

O.O.O.O.Cl[Fe]Cl

InChI

1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

WSSMOXHYUFMBLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Iron(II) chloride tetrahydrate can be used as a catalyst in the:
  • Solvent-free Ritter reaction of nitriles with various halohydrocarbons to synthesize N-substituted amides.
  • Synthesis of ketones via indirect hydration of alkynes.
  • Stereoselective reduction of α, β-unsaturated carbonyl compounds to the corresponding saturated alcohols in the presence of lithium and DTBB.
It can also be used as a precursor to prepare iron-bisphosphine complex, which can be used to catalyze Kumada­-Tamao-­Corriu coupling.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maren Depke et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 85(2), 140-150 (2013-12-19)
Throughout the world, infections caused by bacteria such as Staphylococcus aureus are a major cause of morbidity and mortality. In order to gain some understanding of the complicated physiological link between host and pathogen, modern techniques such as confocal microscopy
Hui Wei et al.
Molecules (Basel, Switzerland), 25(6) (2020-04-05)
Ferrous ion co-catalyst enhancement of dilute-acid (DA) pretreatment of biomass is a promising technology for increasing the release of sugars from recalcitrant lignocellulosic biomass. However, due to the reductive status of ferrous ion and its susceptibility to oxidation with exposure
The crystal structure of iron (II) chloride tetrahydrate.
Penfold BR and Grigor JA.
Acta Crystallographica, 12(11), 850854-850854 (1959)
Electron microscopy study of CVT grown Fe-sulphides.
Zavasnik J and Recnik A.
Journal of Crystal Growth, 367, 18-23 (2013)
Jiangfei Meng et al.
Journal of food science, 77(1), C8-14 (2011-12-21)
Wines made from 3 spine grape (Vitis davidii Foex) genotypes-Junzi 1# (JZ 1#), Junzi 2# (JZ 2#), and Liantang (LT)-and Cherokee rose (Rosa laevigata Michx., CR) were evaluated for their phenolics composition and antioxidant activities by several assays, including 2,2'-azino-bis-(3-ethylbenzothiazoline-6-sulfonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service