Skip to Content
Merck
All Photos(1)

Key Documents

1.02459

Supelco

Iron(II) chloride tetrahydrate

extra pure special grade

Synonym(s):

Ferrous chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2 · 4H2O
CAS Number:
Molecular Weight:
198.81
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes

grade

for analytical purposes

Quality Level

vapor pressure

10 mmHg ( 693 °C)

pH

2.5 (20 °C, 100 g/L in H2O)

mp

107 °C (Elimination of water of crystallisation)

SMILES string

O.O.O.O.Cl[Fe]Cl

InChI

1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

WSSMOXHYUFMBLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service