Skip to Content
Merck
All Photos(1)

Key Documents

31232-M

Sigma-Aldrich

Iron(III) chloride hexahydrate

puriss. p.a., reag. Ph. Eur., ≥99%

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
solid

Agency

USP/NF
reag. Ph. Eur.

Quality Level

vapor pressure

1 mmHg ( 194 °C)

grade

puriss. p.a.

Assay

≥99%

form

solid

impurities

≤0.001% free chlorine (Cl)
≤0.001% total nitrogen (N)
≤0.05% sub.not precip.b.NH4OH(SO4)
≤0.2% free acid (as HCl)

bp

280-285 °C (lit.)

mp

37 °C (lit.)

anion traces

phosphate (PO43-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

As: ≤0.0005%
Cu: ≤0.002%
Fe2+: ≤0.005%
Pb: ≤0.002%
Zn: ≤0.002%

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) chloride hexahydrate can be obtained from crystallization of iron(III) chloride from water. Its crystal structure has been studied using three-dimensional single-crystal X-ray-diffraction data. Its crystals have two chloride ions and four water molecules arranged around each ferric ion to form octahedral trans-[FeCl2(OH2)4]+ ions having C2h-2/m symmetry.

Application

Iron(III) chloride hexahydrate may be employed as an oxidant for the following conversions:
  • Synthesis of 2-substituted 4(3H)-quinazolinones (yield = 77-93%), via condensation of aldehydes with anthranilamide.
  • Oxidation of 1,4-dihydropyridines (1,4-DHPs) to corresponding pyridines.
  • Oxidation of 4-isopropyl-1,4-DHP to dealkylated pyridine.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service