Skip to Content
Merck
All Photos(3)

Key Documents

319996

Sigma-Aldrich

Chlorobenzene

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04

grade

ACS reagent

Quality Level

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

1178 °F

expl. lim.

7.1 %

impurities

≤0.004 meq/g Titr. acid

evapn. residue

≤0.02%

color

APHA: ≤30

refractive index

n20/D 1.524 (lit.)

pH

7

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient recyclable organic solar cells on cellulose nanocrystal substrates with a conducting polymer top electrode deposited by film-transfer lamination.
Zhou Y, et al.
Organic Electronics, 15(3), 661-666 (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 302-302 (1994)
Catalysts based on pillared clays for the oxidation of chlorobenzene.
Aznarez A, et al.
Catalysis Today, 246, 15-27 (2015)
Eagleson M.
Concise Encyclopedia Chemistry, 6-6 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 74-74 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service