Skip to Content
Merck
All Photos(2)

Key Documents

101389

Sigma-Aldrich

Chlorobenzene

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.04

vapor density

3.86 (vs air)

Quality Level

vapor pressure

11.8 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

1178 °F

expl. lim.

7.1 %

refractive index

n20/D 1.524 (lit.)

pH

7

bp

132 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble 0.207 g/L at 20 °C

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chlorobenzene may be used in the synthesis of :
  • DDT (Dichlorodiphenyltrichloroethane)
  • acetophenetidin
  • DL-erythrose derivatives, which allows to convert pollutants to synthetically useful chiral intermediates
  • ortho/para-nitrochlorobenzenes and aniline

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheremisinoff.PN
Industrial Solvents Handbook, Revised And Expanded Chemical Industries, (2) (2003)
"An enantiodivergent approach to D-and L-erythrose via microbial oxidation of chlorobenzene"
Hudlicky T, et al.
Tetrahedron Letters, 30(31), 4053-4054 (1989)
Catalysts based on pillared clays for the oxidation of chlorobenzene.
Aznarez A, et al.
Catalysis Today, 246, 15-27 (2015)
Eagleson M.
Concise Encyclopedia Chemistry, 302-302 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 6-6 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service