Skip to Content
Merck
All Photos(3)

Key Documents

31669

Sigma-Aldrich

Tin(II) chloride dihydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%

Synonym(s):

Stannous chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2 · 2H2O
CAS Number:
Molecular Weight:
225.65
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
form:
fine crystals
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥98%

form

fine crystals

impurities

≤0.002% ammonium (NH4)
≤0.005% insoluble in hydrochloric acid
≤0.01% other heavy metals (as Pb)
≤0.05% subst. not precip. by H2S (SO4)

bp

652 °C (lit.)

mp

37-38 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤20 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤50 mg/kg
Cu: ≤10 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤50 mg/kg

SMILES string

O.O.Cl[SnH2]Cl

InChI

1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2

InChI key

FWPIDFUJEMBDLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tin(II) chloride dihydrate is the hydrated salt of tin(II). Its crystals are monoclinic and their structure has been investigated by single crystal X-ray diffraction method. Cell parameters were reported to be: a=9.313, b=7.250, c=8.970Å, β=114°55′ and Z=4.[1]

Application

Tin(II) chloride dihydrate may be used in the following studies:
  • Synthesis of indoles.[2]
  • Preparation of aldehydes.[3]
  • As Lewis acid catalyst for the synthesis of 3-aminoimidazo[1,2-a]pyridines, via multi-component reactions.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hedieh Saffari et al.
Mayo Clinic proceedings, 95(3), 449-458 (2020-03-07)
To determine if heparin labeled with 99mTechnetium (99mTc) could be an imaging probe to detect eosinophil-related inflammation in eosinophilic esophagitis and to determine the biodistribution and radiation dosimetry of 99mTc-heparin oral administration using image-based dosimetry models with esophageal modeling. Freshly
An X-Ray Redetermination of the Crystal Structure of Tin (II) Chloride Dihydrate.
Kiriyama H, et al.
BuMed News Letter, 46(5), 1389-1395 (1979)
Anupam Bandyopadhyay et al.
Organic & biomolecular chemistry, 8(21), 4855-4860 (2010-08-25)
A facile synthetic route for the preparation of N-protected γ-amino β-keto esters from amino aldehydes and ethyl diazoacetate is described. The two component coupling is facilitated by tin(II) chloride followed by semipinacol rearrangement leading to the product in quantitative yield.
Tin (II) chloride dihydrate: A mild and efficient reagent for cleaving acetals.
Ford KL and Roskamp EJ.
Tetrahedron Letters, 33(9), 1135-1138 (1992)
Ruthenium-catalysed synthesis of indoles from anilines and trialkanolamines in the presence of tin (II) chloride dihydrate.
Cho CS.
Chemical Communications (Cambridge, England), 9, 995-996 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service