Skip to Content
Merck
All Photos(5)

Key Documents

243523

Sigma-Aldrich

Tin(II) chloride dihydrate

ACS reagent, 98%

Synonym(s):

Stannous chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2 · 2H2O
CAS Number:
Molecular Weight:
225.65
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
98%
98.0-103.0% (ACS specification)
grade:
ACS reagent
form:
powder or crystals
solubility:
hydrochloric acid: passes test
water: soluble
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

98%
98.0-103.0% (ACS specification)

form

powder or crystals

reaction suitability

reagent type: catalyst
core: tin

bp

652 °C (lit.)

mp

37-38 °C (dec.) (lit.)

solubility

hydrochloric acid: passes test
water: soluble

anion traces

sulfate (SO42-): passes test (lim. ~0.003%)

cation traces

Ca: ≤0.005%
Fe: ≤0.003%
K: ≤0.005%
Na: ≤0.01%
Pb: ≤0.01%

SMILES string

O.O.Cl[SnH2]Cl

InChI

1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2

InChI key

FWPIDFUJEMBDLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Crystals of SnCl2.2H2O exhibit exhibit monoclinic crystal system and space group P21/c.[1]

Application

Tin(II) chloride dihydrate (SnCl2.2H2O)may be used in the preparation of indoles.[2] It may be used in the transformation of conjugated dioxolones, conjugated and non-conjugated acetals (dimethoxy and diethoxy acetals) to aldehydes.[3]
It plays the role of a catalyst in the synthesis of 3-aminoimidazo[1,2-a]pyridines.[4] It can be used as precursor for the synthesis of tin dioxide (SnO2) nanostructures by thermal degradation at temperature between 400-700 under controlled conditions.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical properties of SnO2 nanostructures prepared via one-step thermal decomposition of tin (II) chloride dihydrate.
Al-Gaashani R, et al.
Materials Science and Engineering, B, 177(6), 462-470 (2012)
Ruthenium-catalysed synthesis of indoles from anilines and trialkanolamines in the presence of tin (II) chloride dihydrate.
Cho CS.
Chemical Communications (Cambridge, England), 9, 995-996 (1998)
An X-Ray Redetermination of the Crystal Structure of Tin (II) Chloride Dihydrate.
Kiriyama H, et al.
Bulletin of the Chemical Society of Japan, 46(5), 1389-1395 (1973)
Tin (II) Chloride Dihydrate Catalyzed Groebke Condensation: An Efficient Protocol for the Synthesis of 3-Aminoimidazo[1,2-a]pyridines.
Shaabani A, et al.
Chin. J. Chem., 27(2), 369-371 (2009)
Tin (II) chloride dihydrate: A mild and efficient reagent for cleaving acetals.
Ford KL and Roskamp EJ.
Tetrahedron Letters, 33(9), 1135-1138 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service