291048
Iodine monochloride solution
1.0 M in methylene chloride
Synonym(s):
Chloroiodide solution, Wijs solution
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
ICl
CAS Number:
Molecular Weight:
162.36
Beilstein:
3587194
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
form:
liquid
Recommended Products
form
liquid
Quality Level
concentration
0.95-1.10 M (by Na2S2O3, titration)
1.0 M in methylene chloride
density
1.42 g/mL at 25 °C
SMILES string
ClI
InChI
1S/ClI/c1-2
InChI key
QZRGKCOWNLSUDK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Polymethylbenzene complexes of iodine and iodine monochloride.
Andrews LJ and Keefer RM.
Journal of the American Chemical Society, 74(18), 4500-4503 (1952)
Ultrasonic-assisted synthesis of flavones by oxidative cyclization of 2'-hydroxychalcones using iodine monochloride.
Lahyani A and Trabelsi M.
Ultrasonics Sonochemistry, 31, 626-630 (2016)
K R Siebenlist et al.
Biochemistry, 39(46), 14171-14175 (2000-11-23)
There are conflicting ideas regarding the location of the carboxyl-terminal regions of cross-linked gamma-chain dimers in double-stranded fibrin fibrils. Some investigators believe that the chains are always oriented longitudinally along each fibril strand and traverse the contacting ends of abutting
Ru-Jin Huang et al.
Analytical chemistry, 81(5), 1777-1783 (2009-02-10)
This study concerns the development of a coupled diffusion denuder system capable of separating and quantifying gaseous molecular iodine (I(2)) and two other highly reactive iodine species, ICl and HOI, which are collectively named activated iodine compounds (AIC). Both I(2)
R M Passos et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 24(8), 787-790 (1991-01-01)
Samples of Schistosoma mansoni soluble adult worm proteins (SWAP) were iodinated with 4-15 mumol I/mg protein using iodine monochloride. The capacity to elicit immediate hypersensitivity reactions of the iodinated derivatives was compared to that of the native SWAP preparations. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service