Skip to Content
Merck
All Photos(1)

Key Documents

714836

Sigma-Aldrich

Iodine monochloride solution

1 M in acetic acid

Synonym(s):

Chloroiodide solution, Wijs solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICl
CAS Number:
Molecular Weight:
162.36
Beilstein:
3587194
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solution

Quality Level

concentration

1 M in acetic acid

density

1.143 g/mL at 25 °C

SMILES string

ClI

InChI

1S/ClI/c1-2

InChI key

QZRGKCOWNLSUDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodine monochloride is an inorganic reagent used in the iodination and chloroiodination of aromatic and unsaturated compounds, respectively. It is also used to cleave carbon-metal bonds.[1]

Application

Iodine monochloride can be used:
  • As a reagent in the preparation of α-iodo β-ketosulfones from β-ketosulfones.[2]
  • In the synthesis of 3-(4-bromo-2-methylphenyl)-4-iodosydnone, which is further used to prepare substituted pyrazoles.[3]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.1 °F

Flash Point(C)

40.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of α-iodo β-ketosulfones and α-iodo methylsulfones using iodine monochloride
Suryakiran N, et al.
Tetrahedron Letters, 47(26), 4319-4323 (2006)
Iodine monochloride
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2013)
Steric effects on the sydnones reactivity. New sydnones and pyrazoles
Dumitracscu F, et al.
ARKIVOC (Gainesville, FL, United States), 2(26), 80-86 (2002)
F Ibrahim et al.
Journal of pharmaceutical and biomedical analysis, 9(2), 101-107 (1991-01-01)
Two simple and sensitive spectrophotometric methods are described for the assay of three MAO inhibitors: isocarboxazid, tranylcypromine sulphate and iproniazid phosphate. The first method is based on the formation of a highly coloured stable radical anion between the drug as
K R Siebenlist et al.
Biochemistry, 39(46), 14171-14175 (2000-11-23)
There are conflicting ideas regarding the location of the carboxyl-terminal regions of cross-linked gamma-chain dimers in double-stranded fibrin fibrils. Some investigators believe that the chains are always oriented longitudinally along each fibril strand and traverse the contacting ends of abutting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service