Skip to Content
Merck
All Photos(3)

Key Documents

208221

Sigma-Aldrich

Iodine monochloride

reagent grade, ≥95%

Synonym(s):

Chloroiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICl
CAS Number:
Molecular Weight:
162.36
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

Assay

≥95%

form

solid or liquid

bp

97.4 °C (lit.)

density

3.24 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClI

InChI

1S/ClI/c1-2

InChI key

QZRGKCOWNLSUDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Iodine monochloride is an interhalogen compound. It forms various complexes with ethyl, isopropyl and t-butylbenzenes. Electrically conducting solution of ICl is obtained on dissolution of ICl in polar solvents. ICl is a green oxidizing agent and participates in the following transformations:
  • aldose hemiacetals to the corresponding aldose lactones
  • diarylmethanols to the corresponding diarylmethanones
  • arylalkylmethanols to the corresponding arylalkylmethanones
  • dialkylmethanols to the corresponding dialkylmethanones

Application

Iodine monochloride (ICl) may be employed as a reagent in the following processes:
  • Halogenation of methoxy and dimethoxybenzenes
  • Synthesis of flavones.
  • Preparation of 1-naphthaldehydes.
It may be used as an iodinating agent in the conversion of alkylboranes to alkyl iodides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1-naphthaldehydes via the cascade reactions of 1-phenylpent-4-yn-2-ols promoted by iodine monochloride
Li B, et al.
Tetrahedron Letters, 57.17, 1843-1846 (2016)
Iodine Monochloride (ICl) as a Highly Efficient, Green Oxidant for the Oxidation of Alcohols to Corresponding Carbonyl Compounds
Wei, Peng, et al.
Synthetic Communications, 45.12, 1457-1470 (2015)
A mild and convenient procedure for conversion of alkenes into alkyl iodides via reaction of iodine monochloride with organoboranes.
Kabalka GW and Gooch III EE.
The Journal of Organic Chemistry, 45(18), 3578-3580 (1980)
Ultrasonic-assisted synthesis of flavones by oxidative cyclization of 2'-hydroxychalcones using iodine monochloride.
Lahyani A and Trabelsi M.
Ultrasonics Sonochemistry, 31, 626-630 (2016)
Polymethylbenzene complexes of iodine and iodine monochloride.
Andrews LJ and Keefer RM.
Journal of the American Chemical Society, 74(18), 4500-4503 (1952)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service