Skip to Content
Merck
All Photos(2)

Key Documents

110280

Sigma-Aldrich

Dibutyl ether

ReagentPlus®, ≥99%

Synonym(s):

Butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2O
CAS Number:
Molecular Weight:
130.23
Beilstein:
1732752
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.48 (vs air)

Quality Level

vapor pressure

4.8 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

365 °F

expl. lim.

8.5 %

refractive index

n20/D 1.399 (lit.)

pH

5.2

bp

142-143 °C (lit.)

mp

−98 °C (lit.)

density

0.764 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCCC

InChI

1S/C8H18O/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

DURPTKYDGMDSBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl ether is commonly used as a solvent for various reactions. It forms azeotropic compositions with various organic solvents and the vapor/liquid equilibria of its binary mixtures have been investigated by head-space gas chromatography.

Application

Dibutyl ether may be used to compose the solvent mixtures in the following studies:
  • To evaluate the anthracene solubility in various binary solvent mixtures composed of dibutyl ether along with n-hexane, cyclohexane, n-heptane, methylcyclohexane, n-octane, isooctane and cyclooctane.
  • To measure the pyrene solubility in various binary solvent mixtures composed of dibutyl ether along with n-hexane, cyclohexane, n-heptane, methylcyclohexane, n-octane, isooctane and tert-butylcyclohexane.
  • To investigate the reaction mechanism and kinetic studies of the reaction of phenyl isocyanate with methanol in the presence of dibutyltin diacetate (catalyst) to afford urethane. Rate constant of the reaction, k was reported to be 0.96liter/(mole sec).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasha A Andrade et al.
Environmental pollution (Barking, Essex : 1987), 222, 412-422 (2017-01-21)
Polybrominated diphenyl ethers (PBDEs) may enter the environment because of accumulation in biosolids followed by application to agricultural lands. No published dissipation studies are available for PBDEs in agricultural soils after biosolids application. Therefore, we conducted a 3-year study to
Solubility of anthracene in binary solvent mixtures containing dibutyl ether.
Marthandan MV and Acree Jr WE.
Journal of Chemical and Engineering Data, 32(3), 301-303 (1987)
Isothermal vapour/liquid equilibria of binary mixtures with dibutyl ether at 298.15 K.
Lepori L, et al.
Physical Chemistry Chemical Physics, 2(21), 4837-4842 (2000)
Solubility of pyrene in binary solvent mixtures containing dibutyl ether.
Wallach JR, et al.
Journal of Chemical and Engineering Data, 34(1), 70-73 (1989)
Kinetics and mechanism of urethane formation catalyzed by organotin compounds. I. The reaction of phenyl isocyanate with methanol in dibutyl ether under the action of dibutyltin diacetate.
Van der Weij FW.
Journal of Polymer Science Part A: Polymer Chemistry, 19(2), 381-388 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service