Skip to Content
Merck
All Photos(3)

Key Documents

271454

Sigma-Aldrich

Dibutyl ether

anhydrous, 99.3%

Synonym(s):

Butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2O
CAS Number:
Molecular Weight:
130.23
Beilstein:
1732752
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor density

4.48 (vs air)

vapor pressure

4.8 mmHg ( 20 °C)

Assay

99.3%

form

liquid

autoignition temp.

365 °F

expl. lim.

8.5 %

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 1.399 (lit.)

pH

5.2

bp

142-143 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: 0.113 g/L at 20 °C

density

0.764 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCCC

InChI

1S/C8H18O/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

DURPTKYDGMDSBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyl ether is being considered as an alternative fuel for diesel engines due to its non-miscibility in water, high cetane number and energy content.
Dibutyl ether is commonly used as solvent for various reactions. It forms azeotropic compositions with various organic solvents and the vapour/liquid equilibria of its binary mixtures have been investigated by head-space gas chromatography.

Application

Dibutyl ether may be used as a solvent for the synthesis of tricarbonyl[(1,2,3,4-η)-1- and 2-methoxy-1,3-cyclohexadiene]iron by the reaction of 1-methoxy-1,4-cyclohexadiene with iron pentacarbonyl.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility of anthracene in binary alcohol+ dibutyl ether solvent mixtures.
Powell JR and Acree Jr.
Journal of Chemical and Engineering Data, 40(4), 914-916 (1995)
A chemical kinetic study of the oxidation of dibutyl-ether in a jet-stirred reactor.
Thion S, et al.
Combustion and Flame, 185, 4-15 (2017)
Seon-Joo Choi et al.
ACS applied materials & interfaces, 10(37), 31404-31412 (2018-08-28)
All-solid-state lithium batteries (ASSLBs) based on sulfide solid electrolytes (SEs) have received great attention because of the high ionic conductivity of the SEs, intrinsic thermal safety, and higher energy density achievable with a Li metal anode. However, studies on practical
Tricarbonyl [(2, 3, 4, 5-?)-2, 4-Cyclohexadien-1-One] Iron and Tricarbonyl [(1, 2, 3, 4, 5-?)-2-Methoxy-2, 4-Cyclohexadien-1-yl] Iron (1+) Hexafluorophosphate (1-) from Anisole.
Birch AJ &amp; Chamberlain KB
Organic Syntheses, 57, 107-107 (1977)
Yangang Ren et al.
The Science of the total environment, 749, 141406-141406 (2020-08-21)
Phytohormones emitted into the atmosphere perform many functions relating to the defence, pollination and competitiveness of plants. To be effective, their atmospheric lifetimes must be sufficient that these signals can be delivered to their numerous recipients. We investigate the atmospheric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service