Skip to Content
Merck
All Photos(1)

Key Documents

Y0001547

Propylene glycol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,2-Propanediol, Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

API family

propylene glycol

autoignition temp.

779 °F

expl. lim.

12.5 %

manufacturer/tradename

EDQM

refractive index

n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Propylene glycol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S LaKind et al.
Critical reviews in toxicology, 29(4), 331-365 (1999-08-18)
The purpose of this article is to review and interpret the scientific literature on the mammalian toxicity of ethylene glycol (EG) and propylene glycol (PG), with the goal of comparing the toxicity of the two chemicals. This type of review
J O Funk et al.
Contact dermatitis, 31(4), 236-241 (1994-10-01)
Evaluation of dermatitis associated with propylene glycol application or ingestion remains a challenge. The research dealing with skin reactions to propylene glycol is revisited and new aspects for future research are outlined. Based on literature review and our own observations
Janina Seubert et al.
PloS one, 9(5), e98347-e98347 (2014-05-31)
Scented cosmetic products are used across cultures as a way to favorably influence one's appearance. While crossmodal effects of odor valence on perceived attractiveness of facial features have been demonstrated experimentally, it is unknown whether they represent a phenomenon specific
Li Wang et al.
Hepatology (Baltimore, Md.), 61(2), 486-496 (2014-09-10)
Defective autophagy is implicated in the pathogenesis of nonalcoholic fatty liver diseases (NAFLD) through poorly defined mechanisms. Cardiolipin is a mitochondrial phospholipid required for bioenergetics and mitophagy from yeast to mammals. Here we investigated a role for ALCAT1 in the
Hongjun Zhang et al.
Stem cells (Dayton, Ohio), 32(8), 2267-2277 (2014-04-10)
Adult stem cells (SCs) are important to maintain homeostasis of tissues including several mini-organs like hair follicles and sweat glands. However, the existence of stem cells in minor salivary glands (SGs) is largely unexplored. In vivo histone2B green fluorescent protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service