Skip to Content
Merck
All Photos(1)

Key Documents

1265515

USP

Ethylene glycol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ethylene glycol, 1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

autoignition temp.

752 °F

expl. lim.

15.3 %

manufacturer/tradename

USP

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethylene glycol USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Propylene Glycol
  • Polyethylene Glycol 3350
  • Noncrystallizing Sorbitol Solution
  • Polyoxyl 40 Hydrogenated Castor Oil
  • Maltitol Solution
  • Sorbitol Solution
  • Sorbitol Sorbitan Solution
  • Dapagliflozin Propanediol

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylene Glycol
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 6164-6164 (2013)
Ethylene Glycol, Diethylene Glycol, and Triethylene Glycol in Ethoxylated Products
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(4), 6733-6733 (2014)
Liang Su et al.
Nature communications, 6, 6287-6287 (2015-02-18)
The enhancement of molecular absorption, emission and scattering processes by coupling to surface plasmon polaritons on metallic nanoparticles is a key issue in plasmonics for applications in (bio)chemical sensing, light harvesting and photocatalysis. Nevertheless, the point spread functions for single-molecule
Peter Mygind Leth et al.
Forensic science international, 155(2-3), 179-184 (2005-10-18)
Ethylene glycol (EG) can be found in many agents, such as antifreeze. Ingestion of EG may cause serious poisoning. Adults are typically exposed when EG is ingested as a cheap substitute for ethanol or in suicide-attempts. Children may be exposed
Eurwin Suryana et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 37, 69-75 (2014-07-11)
Myelination is an essential process in human development that is carried out by oligodendrocytes in the central nervous system. Hypoxic-ischaemic (HI) brain injury can disrupt myelination by causing oxidative stress, inflammation and excitotoxicity, resulting in the loss of myelin as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service