Merck
  • A review of the comparative mammalian toxicity of ethylene glycol and propylene glycol.
A review of the comparative mammalian toxicity of ethylene glycol and propylene glycol.
Critical reviews in toxicology (1999-08-18)
J S LaKind, E A McKenna, R P Hubner, R G Tardiff
ABSTRACT
The purpose of this article is to review and interpret the scientific literature on the mammalian toxicity of ethylene glycol (EG) and propylene glycol (PG), with the goal of comparing the toxicity of the two chemicals. This type of review may serve as the basis for risk management decision-making. Because EG is not a GRAS (generally recognized as safe) chemical, its uses are restricted when compared with PG; thus, certain routes of exposure are not relevant here for toxicological comparison (e.g., subcutaneous, intramuscular, and intravenous). Therefore, this review is focused on the oral, inhalation, and dermal routes of exposure. However, where toxicological data derived from an alternative route of exposure serve to eludicate mechanisms of toxicity, data from these routes are considered. Based on the review provided herein, the following conclusions can be drawn. From the standpoint of lethality, acute effects, and reproductive, developmental, and kidney toxicity, the toxicity of EG exceeds that of PG. Further, localized dermal effects from EG and PG are both mild, with data suggesting that PG may have a skin contact sensitization potential. Finally, PG exposure in laboratory animals has been associated with reversible hematological changes; no data were located for EG from which to draw a toxicological comparison.
Product Description
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%