Skip to Content
Merck
All Photos(1)

Key Documents

Y0001466

Anhydrous Docetaxel

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Docetaxel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H53NO14
CAS Number:
Molecular Weight:
807.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

docetaxel

manufacturer/tradename

EDQM

mp

186-192 °C (dec.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](O)[C@@]3(C)C(=O)[C@H](O)C4=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)c5ccccc5)[C@]3([H])[C@@]1(CO2)OC(C)=O)C4(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c6ccccc6

InChI

1S/C43H53NO14/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45/h9-18,26-28,30-33,35,46-48,53H,19-21H2,1-8H3,(H,44,52)/t26-,27-,28+,30-,31+,32+,33-,35-,41+,42-,43+/m0/s1

InChI key

ZDZOTLJHXYCWBA-VCVYQWHSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Anhydrous Docetaxel EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Docetaxel is a taxane anti-cancer agent related to paclitaxel. It binds to and stabilizes the β−tubulin subunit of microtubules, preventing depolymerization of the mitotic spindle thus leading to cell cycle arrest and apoptosis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Lact. - Muta. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir Goldkorn et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(11), 1136-1142 (2014-03-13)
Circulating tumor cell (CTC) enumeration has not been prospectively validated in standard first-line docetaxel treatment for metastatic castration-resistant prostate cancer. We assessed the prognostic value of CTCs for overall survival (OS) and disease response in S0421, a phase III trial
Jasdeep K Saggar et al.
International journal of cancer, 134(11), 2726-2734 (2013-12-18)
Many chemotherapy drugs have poor therapeutic activity in regions distant from tumor blood vessels because of poor tissue penetration and low cytotoxic activity against slowly-proliferating cells. The hypoxia-activated pro-drug TH-302 may have selective toxicity for hypoxic and neighboring cells in
Edward B Garon et al.
Lancet (London, England), 384(9944), 665-673 (2014-06-17)
Ramucirumab is a human IgG1 monoclonal antibody that targets the extracellular domain of VEGFR-2. We aimed to assess efficacy and safety of treatment with docetaxel plus ramucirumab or placebo as second-line treatment for patients with stage IV non-small-cell-lung cancer (NSCLC)
Monisha Sudarshan et al.
Annals of surgical oncology, 22(1), 324-330 (2014-07-16)
We have previously identified Docetaxel, Cisplatin, and 5FU (DCF) as a safe, tolerable, and effective regimen in the neoadjuvant setting for locally advanced adenocarcinoma (ADC) of the esophagus and esophagogastric junction (EGJ). We hypothesized that DCF combined with enhanced surgical
Alice Dragomir et al.
BMC health services research, 14, 252-252 (2014-06-15)
For Canadian men, prostate cancer (PCa) is the most common cancer and the 3rd leading cause of cancer mortality. Men dying of PCa do so after failing castration. The management of metastatic castration-resistant prostate cancer (mCRPC) is complex and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service