
Anhydrous Docetaxel

Synonym(s):
Docetaxel
Empirical Formula (Hill Notation):
C43H53NO14
CAS Number:
Molecular Weight:
807.88
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing