Skip to Content
Merck
All Photos(1)

Key Documents

PHR1883

Supelco

Docetaxel

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Docetaxel trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H53NO14 · 3H2O
CAS Number:
Molecular Weight:
861.93
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001133
traceable to USP 1224551

API family

docetaxel

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

N([C@H]([C@@H](O)C(=O)O[C@H]2C[C@]3([C@H]([C@@H]5[C@]6([C@H](OC6)C[C@@H]([C@]5(C(=O)[C@@H](C(=C2C)C3(C)C)O)C)O)OC(=O)C)OC(=O)c4ccccc4)O)c1ccccc1)C(=O)OC(C)(C)C.O.O.O

InChI

1S/C43H53NO14.3H2O/c1-22-26(55-37(51)32(48)30(24-15-11-9-12-16-24)44-38(52)58-39(3,4)5)20-43(53)35(56-36(50)25-17-13-10-14-18-25)33-41(8,34(49)31(47)29(22)40(43,6)7)27(46)19-28-42(33,21-54-28)57-23(2)45;;;/h9-18,26-28,30-33,35,46-48,53H,19-21H2,1-8H3,(H,44,52);3*1H2/t26-,27-,28+,30-,31+,32+,33-,35-,41+,42-,43+;;;/m0.../s1

InChI key

XCDIRYDKECHIPE-QHEQPUDQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Docetaxel is a taxoid anticancer agent that binds to β tubulin and promotes tubulin assembly into microtubles inducing cell-cycle arrest and apoptosis.[1]

Application

Docetaxel may be used as a pharmaceutical reference standard for the determination of the analyte in plasma samples by various chromatography techniques.[1][2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB0750 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of docetaxel and ketoconazole in rat plasma by liquid chromatography/electrospray ionization tandem mass spectrometry
Huang Q, et al.
Rapid Communications in Mass Spectrometry, 21(6), 1009-1018 (2007)
Qingguo Zhao et al.
Stem cell research & therapy, 12(1), 29-29 (2021-01-09)
Extracellular vesicles (EVs) and their mimics from mesenchymal stem cells (MSCs) are promising drug carriers to improve cancer treatment, but their application is hindered by donor variations and expansion limitations of conventional tissue-derived MSCs. To circumvent these issues, we made
Sensitive determination of docetaxel in human plasma by liquid--liquid extraction and reversed-phase high-performance liquid chromatography
Loos WJ, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 693(2), 437-441 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service