Skip to Content
Merck
All Photos(1)

Key Documents

Y0000786

Rosmarinic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(R)-O-(3,4-Dihydroxycinnamoyl)-3-(3,4- dihydroxyphenyl)lactic acid, 3,4-Dihydroxycinnamic acid (R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O8
CAS Number:
Molecular Weight:
360.31
Beilstein:
2227587
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rosmarinic acid

manufacturer/tradename

EDQM

mp

171-175 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c(O)c2

InChI

1S/C18H16O8/c19-12-4-1-10(7-14(12)21)3-6-17(23)26-16(18(24)25)9-11-2-5-13(20)15(22)8-11/h1-8,16,19-22H,9H2,(H,24,25)/b6-3+/t16-/m1/s1

InChI key

DOUMFZQKYFQNTF-WUTVXBCWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rosmarinic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Rosmarinic acid has shown to contain antioxidant and anti-inflammatory activities. Possesses promising physiological actions related to cognitive performance, Alzheimer′s disease prevention, kidney disease treatment, cardioprotection and cancer chemoprevention.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luciana Garros Ferreira et al.
Acta cirurgica brasileira, 28 Suppl 1, 83-87 (2013-02-13)
The rationale of the present review is to analize the activity of Rosmarinus officinalis in the the cardiovascular system A MEDLINE database search (from January 1970 to December 2011) using only rosmarinic acid as searched term. The references search revealed
Victor P Bulgakov et al.
Critical reviews in biotechnology, 32(3), 203-217 (2011-08-16)
Rosmarinic acid (RA) is one of the first secondary metabolites produced in plant cell cultures in extremely high yields, up to 19% of the cell dry weight. More complex derivatives of RA, such as rabdosiin and lithospermic acid B, later
Maike Petersen et al.
Phytochemistry, 62(2), 121-125 (2002-12-17)
Rosmarinic acid is an ester of caffeic acid and 3,4-dihydroxyphenyllactic acid. It is commonly found in species of the Boraginaceae and the subfamily Nepetoideae of the Lamiaceae. However, it is also found in species of other higher plant families and
Sarah E Bloch et al.
Chembiochem : a European journal of chemical biology, 15(16), 2393-2401 (2014-09-11)
Hydroxycinnamic acid esters (HCEs) are widely-distributed phenylpropanoid-derived plant natural products. Rosmarinic acid (RA), the most well-known HCE, shows promise as a treatment for cancer and neurological disorders. In contrast to extraction from plant material or plant cell culture, microbial production
Miguel Alcaraz et al.
Anticancer research, 34(4), 1913-1921 (2014-04-03)
Rosmarinic acid is an ester of caffeic acid with interesting biological activities including antioxidant effects and scavenging of oxygen-free radicals. To determine the potentially paradoxical effect of rosmarinic acid, typically being radioprotective when applied to non-tumorous cells, yet conversely displaying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service