Merck
  • Is rosmarinic acid underestimated as an experimental cardiovascular drug?
Is rosmarinic acid underestimated as an experimental cardiovascular drug?
Acta cirurgica brasileira (2013-02-13)
Luciana Garros Ferreira, Andrea Carla Celotto, Verena Kise Capellini, Agnes Afrodite Sumarelli Albuquerque, Tales Rubens de Nadai, Marco Tulio Menezes de Carvalho, Paulo Roberto Barbosa Evora
ABSTRACT
The rationale of the present review is to analize the activity of Rosmarinus officinalis in the the cardiovascular system A MEDLINE database search (from January 1970 to December 2011) using only rosmarinic acid as searched term. The references search revealed 509 references about rosmarinic acid in 40 years (the first reference is from 1970). There is a powerful prevalence of antioxidant and cancer studies. Other diseases are few cited, as inflammation, brain (Alzheimer and Parkinson disease) and, memory; allergy; diabetes; atherosclerosis, and; hypertension. It is necessary to consider the complete absence of studies on coronary artery disease, myocardial ischemia, heart failure or ischemia/reperfusion injury. Rosmarinic acid is underestimated as an experimental cardiovascular drug and deserves more attention.
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Rosmarinic acid, primary reference standard
Rosmarinic acid, European Pharmacopoeia (EP) Reference Standard