Skip to Content
Merck
All Photos(1)

Key Documents

60018

Supelco

Caffeic acid

matrix substance for MALDI-MS, ≥99.0% (HPLC)

Synonym(s):

3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
1954563
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.0% (HPLC)

form

powder

analyte chemical class(es)

peptides, proteins

technique(s)

MALDI-MS: suitable

color

slightly beige

mp

211-213 °C (dec.) (lit.)

cation traces

Ba: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤20 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for UV test

SMILES string

OC(=O)\C=C\c1ccc(O)c(O)c1

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2+

InChI key

QAIPRVGONGVQAS-DUXPYHPUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Caffeic acid is an antioxidant and the major hydroxycinnamic acid present in wine. It contains a catechol group with an α,β-unsaturated carboxylic acid chain with hepatoprotective properties.

Application

Caffeic acid may be used as an electrocatalysis mediator in voltammetric determination of cysteamine (CA) in an aqueous buffer solution.

Biochem/physiol Actions

A natural dietary phenolic compound found in plants that is an anti-oxidant. Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antioxidant activity of caffeic acid (3, 4-dihydroxycinnamic acid).
Gulcin I.
Toxicology, 217(2-3), 213-220 (2006)
Structure-hepatoprotective activity relationship of 3, 4-dihydroxycinnamic acid (caffeic acid) derivatives.
Perez-Alvarez V, et al.
Journal of Applied Toxicology, 21(6), 527-531 (2001)
Electrocatalytic determination of cysteamine using multiwall carbon nanotube paste electrode in the presence of 3, 4-dihydroxycinnamic acid as a homogeneous mediator.
Keyvanfard M, et al.
Journal of the Brazilian Chemical Society, 24(1), 32-39 (2013)
Ren-Wang Jiang et al.
Current medicinal chemistry, 12(2), 237-246 (2005-01-11)
Caffeic acid (3,4-dihydroxycinnamic acid), one of the most common phenolic acids, frequently occurs in fruits, grains and dietary supplements for human consumption as simple esters with quinic acid or saccharides, and are also found in traditional Chinese herbs. Caffeic acid
Morshed A Chowdhury et al.
Journal of medicinal chemistry, 52(6), 1525-1529 (2009-03-20)
A novel class of 1-(4-methanesulfonylphenyl and 4-aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole hybrid cyclooxygenase-2 (COX-2)/5-lipoxygenase (5-LOX) inhibitory anti-inflammatory agents was designed. Replacement of the tolyl ring present in celecoxib by the N-difluoromethyl-1,2-dihydropyrid-2-one moiety provided compounds showing dual selective COX-2/5-LOX inhibitory activities. 1-(4-Aminosulfonylphenyl)-5-[4-(1-difluoromethyl-1,2-dihydropyrid-2-one)]-3-trifluoromethyl-1H-pyrazole exhibited good anti-inflammatory

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service