Skip to Content
Merck
All Photos(1)

Key Documents

PHR1367

Supelco

Sorbic acid

Sorbic acid
1 of 1 reviewers received a sample product or took part in a promotion

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sorbic acid, α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098547
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1615956

vapor pressure

0.01 mmHg ( 20 °C)

API family

sorbic acid

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

132-135 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sorbic acid is a short-chained unsaturated aliphatic carboxylic fatty acid, that is used as an antimicrobial preservative in a wide range of foods, drugs, and cosmetic products.[1][2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Sorbic acid may be used as a pharmaceutical reference standard for the determination of the analyte in raw material, food and pharmaceutical formulations by titrimetry[3] and high performance liquid chromatography.[4][5][6][7][8]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2087 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F

Flash Point(C)

127 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of antimicrobial preservatives in pharmaceutical formulations using reverse-phase liquid chromatography
Radus TP and Gyr G
Journal of Pharmaceutical Sciences, 72(3), 221-224 (1983)
Sorbic acid
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 3622-3622 (2017)
Determination of antimicrobial preservatives in pharmaceutical formulations using reverse-phase liquid chromatography
Radus TP and Gyr G
European Journal of Pharmaceutical Sciences, 72(3), 221-224 (1983)
Optimisation of extraction procedures for analysis of benzoic and sorbic acids in foodstuffs
Mota FJM
Food Chemistry, 82(3), 469-473 (2003)
Wine Analysis and Production
Wine Analysis and Production, 209-215 (1995)

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. West Nyack, NY
    • Reviews 5
    • Vote 1
    5 out of 5 stars.

    the service was great, no issues with the order. thanks

    the service was great, no issues with the order. thanks

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service