Skip to Content
Merck
All Photos(1)

Key Documents

8.22257

Sigma-Aldrich

Benzoic acid

for synthesis

Synonym(s):

Benzoic acid, Phenylformic acid, Benzene carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
UNSPSC Code:
12352106
EC Index Number:
200-618-2
NACRES:
NA.22

vapor pressure

0.001 hPa ( 20 °C)

Quality Level

Assay

≥99% (acidimetric)

form

solid

autoignition temp.

570 °C

potency

1700 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

2.5-3.5 (20 °C in H2O, saturated solution)

bp

249 °C/1013 hPa

mp

121-123 °C

transition temp

flash point 121 °C

density

1.321 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Application

  • Paper Title: A familiar ring to it: biosynthesis of plant benzoic acids - This study explores the biosynthetic pathways leading to benzoic acid in plants, important for understanding plant metabolism and engineering synthetic pathways for production of benzoic acid derivatives (Widhalm & Dudareva, 2015).
  • Paper Title: Pd (II)-catalysed meta-C–H functionalizations of benzoic acid derivatives - Describes a method for the meta-C–H functionalization of benzoic acid derivatives, which could be pivotal for synthetic chemistry applications (Li et al., 2016).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 122 °C
Melting range (upper value): ≤ 125 °C
Identity (IR): passes test

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service