Skip to Content
Merck
All Photos(4)

Key Documents

S1626

Sigma-Aldrich

Sorbic acid

99.0-101.0% anhydrous basis

Synonym(s):

α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098547
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

vapor pressure

0.01 mmHg ( 20 °C)

Quality Level

Assay

99.0-101.0% anhydrous basis

form

(Crystalline powder or granule)

technique(s)

HPLC: suitable

mp

132-135 °C (lit.)

solubility

ethanol: 0.05 g/mL, clear to hazy, colorless to faintly yellow

density

1.2 g/cm3 at 20 °C

antibiotic activity spectrum

fungi

lipid type

unsaturated FAs

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sorbic acid is used as a food preservative and has antimicrobial property.

Application

Sorbic acid has been used as an antimicrobial component of artificial diet for worms.

Biochem/physiol Actions

Sorbic acid can be used to inhibit bacterial, yeast and fungal sulfhydryl enzymes by inhibiting amino acid uptake.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diet improvement for western corn rootworm (Coleoptera: Chrysomelidae) larvae
Huynh MP, et al.
PLoS ONE, 12(11), e0187997-e0187997 (2017)
Evidence that sorbic acid does not inhibit yeast as a classic `weak acid preservative?
Stratford M and Anslow PA
Letters in Applied Microbiology, 27(4), 203-206 (1998)
Evidence that sorbic acid does not inhibit yeast as a classic `weak acid preservative?
Introduction to Food Toxicology (2009)
A technique for continuous mass rearing of the black vine weevil, Otiorhynchus sulcatus
Fisher JR and Bruck DJ
Entomologia Experimentalis et Applicata, 113(1), 71-75 (2004)
SYNERGY OF SECONDARY COMPOUNDS IN THE ARTIFICIAL FOODS OF THE LAST INSTAR OF HYPHANTRIA CUNEA (DRURY)(LEPIDOPTERA: ARCTIIDAE)
Yanar O, et al.
APPLIED ECOLOGY AND ENVIRONMENTAL RESEARCH, 14(3), 195-205 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service