Skip to Content
Merck
All Photos(1)

Key Documents

PHR1319

Supelco

Residual Solvent - Trichloroethylene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Trichloroethylene, TCE, Trichloroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH=CCl2
CAS Number:
Molecular Weight:
131.39
Beilstein:
1736782
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1601827

vapor density

4.5 (vs air)

vapor pressure

61 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

770 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.476 (lit.)

bp

86.7 °C (lit.)

mp

−84.8 °C (lit.)

density

1.463 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-30°C

SMILES string

Cl\C=C(\Cl)Cl

InChI

1S/C2HCl3/c3-1-2(4)5/h1H

InChI key

XSTXAVWGXDQKEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Residual Solvent - Trichloroethylene belongs to the group of class 2 residual solvents. The use of class 2 solvents should be limited in drug substances, excipients, dietary ingredients, and official products because of the inherent toxicities of these residual solvents. Trichloroethylene is a volatile and chlorinated organic compound, which finds applications as a solvent and greasing agent.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0678 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hideaki Watanabe
The Journal of dermatology, 38(3), 229-235 (2011-02-24)
The number of patients suffering from trichloroethylene (TCE)-related severe skin disorders with liver dysfunction has been increasing in developing countries in Asia, especially since the mid-1990s. In Japan, five cases of this disease have been reported, but none since the
Yih-Horng Shiao
Environmental and molecular mutagenesis, 50(1), 68-77 (2008-11-26)
Trichloroethylene (TCE), an organic solvent commonly used for metal degreasing and as a chemical additive, is a significant environmental contaminant that poses health concerns in humans. The US Environmental Protection Agency (EPA) is currently revising the 2001 TCE human risk
Berit Bakke et al.
Journal of occupational and environmental hygiene, 4(5), 375-390 (2007-04-25)
This article describes a systematic review of the industrial hygiene literature for uses of trichloroethylene (TCE) in industry for the exposure assessment of two population-based case control studies of brain cancer in the United States. Papers and reports that address
Jane C Caldwell et al.
Environmental and molecular mutagenesis, 49(2), 142-154 (2007-11-02)
The mode(s) of action (MOA) of a pollutant for adverse health effects may be dependent on the mixture of metabolites resulting from exposure to a single agent and may also be affected by coexposure to pollutants that have similar targets
Cheryl Siegel Scott et al.
International journal of environmental research and public health, 8(11), 4238-4272 (2011-12-14)
We conducted a meta-analysis focusing on studies with high potential for trichloroethylene (TCE) exposure to provide quantitative evaluations of the evidence for associations between TCE exposure and kidney, liver, and non-Hodgkin lymphoma (NHL) cancers. A systematic review documenting essential design

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service