Skip to Content
Merck
All Photos(1)

Key Documents

PHR1033

Supelco

Niacinamide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Nicotinamide, Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein:
383619
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 460
traceable to Ph. Eur. N0600000
traceable to USP 1462006

API family

nicotinamide

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

128-131 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Niacinamide may be used as a pharmaceutical reference standard for the determination of the analyte in plasma, pharmaceutical formulations and infant/Adult nutritional formula powder by various chromatography methods.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor
Niacinamide/Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1697 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of water-soluble vitamins in SRM 1849 Infant/Adult Nutritional Formula powder by liquid chromatography--isotope dilution mass spectrometry
Goldschmidt RJ and Wolf WR
Analytical and Bioanalytical Chemistry, 397(2), 471-481 (2010)
Rapid determination of thiamine, riboflavin, niacinamide, pantothenic acid, pyridoxine, folic acid and ascorbic acid in Vitamins with Minerals Tablets by high-performance liquid chromatography with diode array detector
Jin P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 70(5), 151-157 (2012)
P Pfuhl et al.
Journal of pharmaceutical and biomedical analysis, 36(5), 1045-1052 (2004-12-29)
A sensitive, specific, accurate, and reproducible HPLC/MS-method for the simultaneous quantitative determination of niacin (NA) and its main metabolites niacinamide (NAM) and nicotinuric acid (NUR) in human plasma using chinolin-3-carboxylic acid as an internal standard was developed and validated according
Mitchell E Horwitz et al.
The Journal of clinical investigation, 124(7), 3121-3128 (2014-06-10)
Delayed hematopoietic recovery is a major drawback of umbilical cord blood (UCB) transplantation. Transplantation of ex vivo-expanded UCB shortens time to hematopoietic recovery, but long-term, robust engraftment by the expanded unit has yet to be demonstrated. We tested the hypothesis
Salvatore Simmini et al.
Nature communications, 5, 5728-5728 (2014-12-17)
The endodermal lining of the adult gastro-intestinal tract harbours stem cells that are responsible for the day-to-day regeneration of the epithelium. Stem cells residing in the pyloric glands of the stomach and in the small intestinal crypts differ in their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service