Skip to Content
Merck
All Photos(1)

Key Documents

PHR1380

Supelco

Ivermectin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ivermectin, 22,23-Dihydroavermectin B1, MK-933

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 864
traceable to Ph. Eur. I8000010
traceable to USP 1354309

API family

ivermectin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]1(O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C6/CO[C@]7([H])[C@H](O)C(C)=C[C@@H](C(=O)O3)[C@]67O)O2)[C@@H](C)CC

InChI

1S/C48H74O14/c1-11-25(2)43-28(5)17-18-47(62-43)23-34-20-33(61-47)16-15-27(4)42(26(3)13-12-14-32-24-55-45-40(49)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-10)44(31(8)57-39)60-38-21-36(53-9)41(50)30(7)56-38/h12-15,19,25-26,28,30-31,33-45,49-50,52H,11,16-18,20-24H2,1-10H3/b13-12+,27-15+,32-14+/t25-,26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41-,42-,43+,44-,45+,47+,48+/m0/s1

InChI key

AZSNMRSAGSSBNP-XPNPUAGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Ivermectin is a derivative of avermectin B1 that is used against both endo- and ectoparasites in veterinary applications. It has also been indicated as a potential drug for the management of Onchocerca volvulus, identified as the cause of ′river blindness′ in humans.

Application

Ivermectin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and plasma samples by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1636 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivermectin (3.15%) long-acting formulations in cattle: absorption pattern and pharmacokinetic considerations
Lifschitz A, et al.
Veterinary Parasitology, 147(3-4), 303-310 (2007)
Ivermectin disposition kinetics after subcutaneous and intramuscular administration of an oil-based formulation to cattle
Lifschitz A, et al.
Veterinary Parasitology, 86(3), 203-215 (1999)
Feng Xiong et al.
The Plant cell, 32(12), 3812-3824 (2020-09-30)
Plant roots are sustained through meristem activity at the root tip. Two transcriptional pathways, one mediated by PLETHORAs (PLTs) and the other by SHORTROOT and SCARECROW, play major roles in root meristem development. The role of PLTs during root meristem
Liquid chromatographic determination of ivermectin in animal plasma with trifluoroacetic anhydride and N-methylimidazole as the derivatization reagent
de Montigny P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 8(6), 507-511 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service