Skip to Content
Merck
All Photos(2)

Key Documents

72345

Sigma-Aldrich

Nicotinamide

≥98.5% (HPLC)

Synonym(s):

Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein:
383619
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥98.5% (HPLC)

form

powder

color

colorless to white

mp

128-131 °C (lit.)
128-131 °C

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinamide is synthesized in the liver from nicotinic acid. It is also produced as a result of nicotinamide adenine dinucleotide (NAD+) hydrolysis.

Application

Nicotinamide has been used:
  • as a precursor to study its effect on nicotinamide adenine dinucleotide (NAD) concentration in NAD-deficient mammalian mitochondria
  • as an inhibitor of sirtuin (SIRT) family of deacetylases in in vitro serine hydroxymethyltransferase 2 (SHMT2) deacetylation assay
  • to study its effect on ischemia-reperfusion (I-R) injury-induced cell death in ptenαm/m mice

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor
Nicotinamide serves as a precursor for β-nicotinamide adenine dinucleotide (NAD+) coenzyme production. It is involved in the synthesis of NAD+ phosphate (NADP+). It also regulates the mechanism of cell survival and cell death. Fatigue, skin pigmentation, decreased appetite and mouth ulcers are indications of nicotinamide deficiency. Its chronic deficiency can lead to pellagra. Nicotinamide is known to show protective effects on oxidative stress.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTEN alpha regulates mitophagy and maintains mitochondrial quality control
Li G, et al.
Autophagy, 14(10), 1742-1760 (2018)
Rūta Barisevičiūtė et al.
PloS one, 15(8), e0237605-e0237605 (2020-08-18)
The vertical distribution of radiocarbon (14C) was examined in the bottom sediment core, taken from Lake Drūkšiai, which has served as a cooling pond since 1983 for the 26 years of the Ignalina Nuclear Power Plant (INPP) operation using two
Nicotinamide adenine dinucleotide is transported into mammalian mitochondria
Davila A, et al.
eLife, 7(9), e33246-e33246 (2018)
Deacetylation of serine hydroxymethyl-transferase 2 by SIRT3 promotes colorectal carcinogenesis
Wei Z, et al.
Nature Communications, 9(1), 4468-4468 (2018)
William J Quinn et al.
Cell reports, 33(11), 108500-108500 (2020-12-17)
Immune cell function is influenced by metabolic conditions. Low-glucose, high-lactate environments, such as the placenta, gastrointestinal tract, and the tumor microenvironment, are immunosuppressive, especially for glycolysis-dependent effector T cells. We report that nicotinamide adenine dinucleotide (NAD+), which is reduced to NADH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service