Merck
  • Transformation of intestinal stem cells into gastric stem cells on loss of transcription factor Cdx2.
Transformation of intestinal stem cells into gastric stem cells on loss of transcription factor Cdx2.
Nature communications (2014-12-17)
Salvatore Simmini, Monika Bialecka, Meritxell Huch, Lennart Kester, Marc van de Wetering, Toshiro Sato, Felix Beck, Alexander van Oudenaarden, Hans Clevers, Jacqueline Deschamps
ABSTRACT
The endodermal lining of the adult gastro-intestinal tract harbours stem cells that are responsible for the day-to-day regeneration of the epithelium. Stem cells residing in the pyloric glands of the stomach and in the small intestinal crypts differ in their differentiation programme and in the gene repertoire that they express. Both types of stem cells have been shown to grow from single cells into 3D structures (organoids) in vitro. We show that single adult Lgr5-positive stem cells, isolated from small intestinal organoids, require Cdx2 to maintain their intestinal identity and are converted cell-autonomously into pyloric stem cells in the absence of this transcription factor. Clonal descendants of Cdx2(null) small intestinal stem cells enter the gastric differentiation program instead of producing intestinal derivatives. We show that the intestinal genetic programme is critically dependent on the single transcription factor encoding gene Cdx2.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard