Skip to Content
Merck
All Photos(1)

Key Documents

P3100000

Procaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Aminobenzoic acid 2-diethylaminoethyl ester, p-Aminobenzoic acid diethylaminoethyl ester hydrochloride, Novocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2CH2CH2N(C2H5)2·HCl
CAS Number:
Molecular Weight:
272.77
Beilstein:
3917802
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

procaine

manufacturer/tradename

EDQM

mp

155-156 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CCN(CC)CCOC(=O)c1ccc(N)cc1

InChI

1S/C13H20N2O2.ClH/c1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h5-8H,3-4,9-10,14H2,1-2H3;1H

InChI key

HCBIBCJNVBAKAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Procaine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oktay Korun et al.
Artificial organs, 37(1), 76-81 (2013-01-12)
Cardioplegic arrest is one of the most common myocardial protection strategies. A wide variety of cardioplegic solutions are routinely being used. There is an ongoing discussion about the relative effectiveness of these solutions considering myocardial protection. This study aims to
Qinlong Liu et al.
Langenbeck's archives of surgery, 397(8), 1323-1331 (2012-11-01)
Ischemia/reperfusion injury is an obstacle especially in steatotic livers, including those with steatosis induced by acute toxic stress. Recently, a modified histidine-tryptophan-ketoglutarate (HTK) solution, HTK-N, has been developed. This solution contains N-acetylhistidine, amino acids, and iron chelators. This study was
Christopher M Barbieri et al.
Journal of molecular biology, 425(10), 1612-1626 (2013-02-13)
The OmpR/PhoB family of response regulators (RRs) is the largest class of two-component system signal transduction proteins. Extensive biochemical and structural characterization of these transcription factors has provided insights into their activation and DNA-binding mechanisms. For the most part, OmpR/PhoB
Jitka Siroká et al.
Journal of pharmaceutical and biomedical analysis, 84, 140-147 (2013-07-09)
A simple capillary electrophoretic method with spectrophotometric UV detection at 236 nm has been developed for the selective separation and determination of 1-(2-chlorophenyl)piperazine (oCPP), 1-(3-chlorophenyl)piperazine (mCPP) and 1-(4-chlorophenyl)piperazine (pCPP) in confiscated pills. Several cyclodextrin derivatives were tested to compose the
Wataru Nakanishi et al.
PloS one, 7(8), e42319-e42319 (2012-08-18)
Several studies have reported that pancreatic ductal preservation greatly improved the islet yield and function after cold storage. However, these studies were devoid of appropriate controls, such as vascular perfusion, which is routinely performed to preserve organs in the clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service