Merck
  • Ductal injection does not increase the islet yield or function after cold storage in a vascular perfusion model.
Ductal injection does not increase the islet yield or function after cold storage in a vascular perfusion model.
PloS one (2012-08-18)
Wataru Nakanishi, Takehiro Imura, Akiko Inagaki, Yasuhiro Nakamura, Satoshi Sekiguchi, Keisei Fujimori, Susumu Satomi, Masafumi Goto
ABSTRACT
Several studies have reported that pancreatic ductal preservation greatly improved the islet yield and function after cold storage. However, these studies were devoid of appropriate controls, such as vascular perfusion, which is routinely performed to preserve organs in the clinical setting. In this study, we created a vascular perfusion model using inbred rats, and investigated the effect of ductal injection on the islet yield and function after cold storage. Rat pancreases after 10 h cold ischemia were classified as follows: without ductal/vascular perfusion; with ductal injection; with vascular perfusion; and with ductal/vascular perfusion. The islet yield, function, viability, release of inflammatory mediators, and pathological changes in the exocrine tissues were assessed in the Hanks' Balanced Salt Solution (HBSS) model. The islet yield was also assesed by introducing University of Wisconsin Solution (UWS) and Histidine-Tryptophan-Ketoglutarate solution (HTK), which are the standard clinical preservation solutions. In the HBSS model, ductal injection and vascular perfusion significantly improved the islet yield compared with the control group. However, ductal injection showed no additional effects on the islet yield, function, viability and suppressing the release of inflammatory mediators when vascular perfusion was performed. Although ductal injection significantly decreased the apoptosis of exocrine cells, no beneficial effect on vacuolation was observed. In contrast, vascular perfusion significantly suppressed vacuolation in the exocrine tissues. Likewise, in the UWS and HTK model, ductal injection and vascular perfusion improved the islet yield compared with the control group. Nevertheless, the combination group showed no additional effects. These data suggest that ductal injection has no additional effect on islet yield and function after cold storage in a vascular perfusion model. We propose that ductal injection can be an effective and simple alternative for vascular perfusion prior to pancreas harvest, but is not necessary in most cases, since vascular perfusion is routinely performed.
Product Description
Supelco
Procaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Procaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Procaine hydrochloride, ≥97%