Skip to Content
Merck
All Photos(1)

Key Documents

M2300000

Minoxidil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

6-(1-Piperidinyl)-2,4-pyrimidinediamine 3-oxide, 6-(1-Piperidinyl)pyrimidine-2,4-diamine 3-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N5O
CAS Number:
Molecular Weight:
209.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

minoxidil

manufacturer/tradename

EDQM

mp

272-274 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC1=CC(=NC(=N)N1O)N2CCCCC2

InChI

1S/C9H15N5O/c10-7-6-8(12-9(11)14(7)15)13-4-2-1-3-5-13/h6,11,15H,1-5,10H2

InChI key

ZIMGGGWCDYVHOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Minoxidil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Activates ATP-activated K+ channels; vasodilator; slow or stop hair loss and promote hair regrowth.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chester L Bowen et al.
Bioanalysis, 4(23), 2823-2832 (2012-12-12)
Stabilization of phase II metabolites is an important consideration during bioanalytical method development, method validation and sample analysis. Generic approaches to stabilize these metabolites during storage in liquid-based matrices include pH adjustment of samples prior to storage and/or temperature control;
Natasha Atanaskova Mesinkovska et al.
Dermatologic clinics, 31(1), 119-127 (2012-11-20)
Female pattern hair loss (FPHL) is the most common cause of alopecia in women. FPHL is characterized histologically with increased numbers of miniaturized, velluslike hair follicles. The goal of treatment of FPHL is to arrest hair loss progression and stimulate
R L DeVillez
Dermatologic clinics, 8(2), 367-375 (1990-04-01)
Topical minoxidil is a trichogenic agent that stimulates the hair follicle via the vasoactive metabolite minoxidil sulfate without any evidence of antiandrogen activity or an effect on the immune system. Less than 5% of the applied dose is absorbed. The
Guilherme Martins Gelfuso et al.
Journal of pharmaceutical sciences, 102(5), 1488-1494 (2013-03-02)
Although minoxidil (MX) is a drug known to stimulate hair growth, the treatment of androgenic alopecia could be improved by delivery strategies that would favor drug accumulation into the hair follicles. This work investigated in vitro the potential of iontophoresis
S E Whitmore
Archives of dermatology, 128(5), 653-656 (1992-05-01)
The correct selection of vehicles for patch testing specific substances is essential in the evaluation of suspected allergic contact dermatitis. A reaction to a chemical may be dependent not only on the concentration tested but also on the vehicle in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service