Skip to Content
Merck
All Photos(3)

Key Documents

P2759

Sigma-Aldrich

Phloxine B

Powder

Synonym(s):

2′,4′,5′,7′-Tetrabromo-4,5,6,7-tetrachlorofluorescein disodium salt, Acid Red 92, Cyanosine, D&C;Red No. 28, Eosin 10B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H2Br4Cl4Na2O5
CAS Number:
Molecular Weight:
829.63
Colour Index Number:
45410
Beilstein:
3887100
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Phloxine B, antibacterial fluorescent dye

Quality Level

form

powder

technique(s)

microbe id | staining: suitable

color

dark red

solubility

water: 1 mg/mL, clear, red

εmax

1150-1300 at 546-550 nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]c1c(Br)cc2c(Oc3c(Br)c([O-])c(Br)cc3C24OC(=O)c5c(Cl)c(Cl)c(Cl)c(Cl)c45)c1Br

InChI

1S/C20H4Br4Cl4O5.2Na/c21-5-1-3-17(9(23)15(5)29)32-18-4(2-6(22)16(30)10(18)24)20(3)8-7(19(31)33-20)11(25)13(27)14(28)12(8)26;;/h1-2,29-30H;;/q;2*+1/p-2

InChI key

OOYIOIOOWUGAHD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Phloxine B has been used to trace the uptake of double stranded RNA by green peach aphids. It has also been used to stain yeast strains.

Biochem/physiol Actions

Phloxine B has been demonstrated as an alternative to Gram staining. It was found to inhibit the growth of methicillin-resistant Staphylococcus aureus. Phloxine B stimulated and inhibited the channel activity of wild-type cystic fibrosis transmembrane conductance regulator (CFTR). It has antibacterial properties. Phloxine B staining can be used to distinguish between gram-positive and gram-negative bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using Vital Dyes to Trace Uptake of dsRNA by Green Peach Aphid Allows Effective Assessment of Target Gene Knockdown
International Journal of Molecular Sciences (2017)
Phloxine B, a versatile bacterial stain
Reuven Rrasooly
Pathogens and Disease (2007)
Advances in measuring lifespan in the yeast Saccharomyces cerevisiae
Nadege Minios
Proceedings of the National Academy of Sciences of the USA (2004)
J R Walton
Neurotoxicology, 30(6), 1059-1069 (2009-07-14)
A recent longitudinal study described an inducible rodent model for age-related cognitive deterioration. This model was produced by chronically feeding rats aluminum, from age 12 months onwards, in measured amounts equivalent to total aluminum levels ingested by Americans from their
Maged Fouad et al.
Electrophoresis, 29(11), 2280-2287 (2008-05-01)
We describe a new and selective analytical method for the separation and quantitation of plant glucosinolates. The new method, which utilizes microchip CE (micro-CE) with fluorescence detection, circumvents the multistep procedures characteristic of conventional methods. Glucosinolates form charge transfer complexes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service