Skip to Content
Merck
All Photos(3)

Key Documents

H3136

Sigma-Aldrich

Hematoxylin

certified by the Biological Stain Commission

Synonym(s):

Natural Black 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O6 · xH2O
CAS Number:
Molecular Weight:
302.28 (anhydrous basis)
Colour Index Number:
75290
Beilstein:
91400
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

mp

200 °C (dec.) (lit.)

solubility

ethanol: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Hematoxylin is a basophilic dye extracted from the tree Heamatoxylum campechianum. It specifically binds to acidic compounds and thus is useful for staining cell nuclei which contain acidic substances: DNA and RNA. Hematoxylin, when oxidized forms hematein, which has thick pale blue purple colour. Hematoxylin is often used along with eosin, in histological study under light microscope.

Application

Hematoxylin has been used:
  • to visualize anatomical features and to detect nucleus locus coeruleus adjacent to the periaqueductal gray region in brainstem samples
  • as a nuclear counterstain to visualize the nucleus, adipocytes were counterstained with a hematoxylin reagent
  • to stain lung tissue for histological analysis

Suitability

Certified for use in Weigert′s iron hematoxylin, Mayer′s hemalum, Harris hemalum and in Heidenhain′s technique

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of matrilin-2 in liver cirrhosis and hepatocellular carcinoma
Szabo E, et al.
Pathology Oncology Research, 14(1), 15-22 (2008)
Histologic validation of locus coeruleus MRI contrast in post-mortem tissue.
Keren NI
Neuroimage, 113, 235-235 (2015)
Techniques in Aquatic Toxicology (2005)
S-nitrosylation of fatty acid synthase regulates its activity through dimerization.
Choi MS
Journal of Lipid Research, 57, 607-607 (2016)
Expression of matrilin-2 in liver cirrhosis and hepatocellular carcinoma.
Szabo E
Pathology Oncology Research, 14, 15-15 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service