Skip to Content
Merck
All Photos(1)

Key Documents

BCR269

Chrysene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909297
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

448 °C (lit.)

mp

252-254 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3c4ccccc4ccc23

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
3575494035769215
Chrysene BCR®, certified reference material

BCR269

Chrysene

Chrysene analytical standard

35754

Chrysene

Chrysene certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

94035

Chrysene

Chrysene

769215

Chrysene

format

neat

format

neat

format

neat

format

-

grade

certified reference material

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

-

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-

manufacturer/tradename

JRC

manufacturer/tradename

-

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

manufacturer/tradename

-

agency

BCR®

agency

-

agency

-

agency

-

Analysis Note

For more information please see:
BCR269

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Theodoros Tzamkiozis et al.
    Inhalation toxicology, 22 Suppl 2, 59-69 (2010-10-30)
    This study presents different research techniques linked together to improve our understanding of the particulate matter (PM) impacts on health. PM samples from the exhaust of different vehicles were collected by a versatile aerosol concentration enrichment system (VACES). Waterborne PM
    Bimal K Banik et al.
    European journal of medicinal chemistry, 45(10), 4687-4691 (2010-08-13)
    We describe herein the synthesis of novel 6,12-distributed chrysene as potent anticancer agents. In vitro and in vivo studies are also reported here.
    José M Lorenzo et al.
    Meat science, 89(1), 105-109 (2011-04-20)
    The polycyclic aromatic hydrocarbon (PAH) content in two traditional smoked sausages from Spain was determined. Results showed, that total average levels of PAHs found were higher in "Chorizo de cebolla" (101.81 vs. 98.48 μgkg(-1)). Most contents of PAHs found in
    L Viñas et al.
    Chemosphere, 75(4), 534-541 (2009-01-20)
    Aliphatic and aromatic hydrocarbons were determined in three species of commercial shellfish, namely razor shells (Ensis arcuatus and Ensis siliqua), goose barnacle (Pollicipes cornucopia) and sea urchin (Paracentrotus lividus), living in different habitats and exhibiting different feeding behaviors. The samples
    M J Smith et al.
    Journal of hazardous materials, 192(3), 1219-1225 (2011-07-12)
    A comparison was made between the dissipation of polycyclic aromatic hydrocarbons (PAHs) in soil freshly spiked with pure PAHs, soil spiked with a coal tar mixture and a contaminated soil from a former coking works where the PAHs had been

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service