Skip to Content
Merck
All Photos(1)

Key Documents

769215

Sigma-Aldrich

Chrysene

Synonym(s):

1,2-Benzophenanthrene, Phenacene, Benzo[a]phenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909297
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

bp

448 °C (lit.)

mp

252-254 °C (lit.)

SMILES string

c1ccc2c(c1)ccc3c4ccccc4ccc23

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H

InChI key

WDECIBYCCFPHNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Chrysene can be used as a building block to synthesize:
  • Chrysene-based tetracarboxy-substituted bis[5]helicenes via double Perkin reactions followed by Pd-catalyzed cyclizations.
  • Chrysene-based azahelicene derivatives are applicable in Perovskite solar cells.
  • 3,6,9,12-tetrakis(4-tert-butylphenyl)chrysene applicable as a blue emitter for OLEDs.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex S Ionkin et al.
Chemical communications (Cambridge, England), (20), 2319-2321 (2008-05-14)
The first tetra-substituted non-fused chrysene, 3,6,9,12-tetrakis(4-tert-butylphenyl)chrysene with blue electroluminescence at 450 nm, and with a radiance of 500 cd m(-2), was synthesized by a two-step procedure: direct bromination of chrysene in trimethyl phosphate, followed by palladium-catalyzed cross-coupling of tetrabromochrysene and
Zefeng Tang et al.
ChemSusChem, 14(22), 4923-4928 (2021-10-13)
Chrysene is a readily available material for exploring new polycyclic aromatic hydrocarbons (PAHs). In this study, two chrysene based azahelicenes, nine-membered BA7 and ten-membered DA6, are constructed by intermolecular oxidative annulation of 6-aminochrysene and intramolecular annulation of N6 ,N12 -bis(1-chloronaphthalen-2-yl)chrysene-6,12-diamine
From chrysene to double [5] Helicenes
Bock H, et al.
European Journal of Organic Chemistry, 2015(5), 1033-1039 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service