Merck
  • Polycyclic aromatic hydrocarbons (PAHs) in two Spanish traditional smoked sausage varieties: "Chorizo gallego" and "Chorizo de cebolla".
Polycyclic aromatic hydrocarbons (PAHs) in two Spanish traditional smoked sausage varieties: "Chorizo gallego" and "Chorizo de cebolla".
Meat science (2011-04-20)
José M Lorenzo, Laura Purriños, Roberto Bermudez, Noemí Cobas, Maria Figueiredo, María C García Fontán
ABSTRACT
The polycyclic aromatic hydrocarbon (PAH) content in two traditional smoked sausages from Spain was determined. Results showed, that total average levels of PAHs found were higher in "Chorizo de cebolla" (101.81 vs. 98.48 μgkg(-1)). Most contents of PAHs found in both sausages were phenanthrene, naphthalene and anthracene. The maximum level for benzo[a]pyrene (BaP) of 5 μgkg(-1) in smoked meat products was not exceeded in any samples. BaP represented a 0.73% and 1.15% with respect to the total sum of the 15 PAHs that were investigated in this study for "Chorizo gallego" and "Chorizo de cebolla", respectively. Correlation statistic analysis (P < 0.01) of the contents from samples analysed, showed that BaP was a good marker for Σ15 PAHs in "Chorizo gallego" samples (R(BaP/Σ15PAHs) = 0.77; P < 0.01) and it was a good marker for 7 U.S. EPA probably carcinogenic PAHs in "Chorizo de cebolla" samples (R(BaP/7EPA) = 0.95) and in "Chorizo gallego" samples (R(BaP/7EPA) = 0.81).
Product Description
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chrysene, analytical standard
Chrysene, BCR®, certified reference material